Skip to Content
Merck
All Photos(1)

Documents

08062601

F11 cell line from rat

Synonym(s):

F-11 Cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
NACRES:
NA.81

form

liquid

Quality Level

shipped in

dry ice

storage temp.

−196°C

General description

For complete product information, please see ECACC

Cell Line Origin

A somatic cell hybrid of a rat embryonic dorsal root ganglion (DRG) and mouse neuroblastoma cell line N18TG2. The line retains both rat and mouse chromosomes and synthesises both rat and mouse isoenzymes. Manifests a unique combination of action potential properties and cell surface markers.

Application

Recommended culture medium: DMEM + 2mM Glutamine + 10% FBS

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service