Skip to Content
Merck
All Photos(1)

Key Documents

06665

Sigma-Aldrich

(S)-trans-2-Amino-4-(2-aminoethoxy)-3-butenoic acid hydrochloride

≥93% (AT)

Synonym(s):

L-α-(2-Aminoethoxyvinyl)glycine hydrochloride, AVG-HCl

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H12N2O3 · HCl
CAS Number:
Molecular Weight:
196.63
Beilstein:
6715149
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.77

Assay

≥93% (AT)

form

powder

optical activity

[α]/D +83±5°, c = 1% in H2O

storage temp.

−20°C

SMILES string

Cl[H].NCCO\C=C\[C@H](N)C(O)=O

InChI

1S/C6H12N2O3.ClH/c7-2-4-11-3-1-5(8)6(9)10;/h1,3,5H,2,4,7-8H2,(H,9,10);1H/b3-1+;/t5-;/m0./s1

InChI key

ZDCPLYVEFATMJF-BTIOQYSDSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Inhibition of ethylene biosynthesis delays ripening of fruits in plants treated with AVG.

Other Notes

AVG is a potent inhibitor of ethylene synthesis in plants acting at the level of 1-aminocyclopropanecarboxylic acid synthase

Disclaimer

The product is not intended for use as a biocide under global biocide regulations, including but not limited to US EPA′s Federal Insecticide Fungicide and Rodenticide Act, European Biocidal Products Regulation, Canada’s Pest Management Regulatory Agency, Turkey’s Biocidal Products Regulation, Korea’s Consumer Chemical Products and Biocide Safety Management Act (K-BPR) and others.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ruifeng Han et al.
PloS one, 13(9), e0204488-e0204488 (2018-09-22)
Nitrogen (N) supply, including NO3--N and organic N in the form of amino acids can influence the morphological attributes of plants. For example, amino acids contribute to plant nutrition; however, the effects of exogenous amino acids on NO3--N uptake and
Y B Yu et al.
Plant physiology, 64(6), 1074-1077 (1979-12-01)
Auxin is known to stimulate greatly both C(2)H(4) production and the conversion of methionine to ethylene in vegetative tissues, while amino-ethoxyvinylglycine (AVG) or Co(2+) ion effectively block these processes. To identify the step in the ethylene biosynthetic pathway at which
Chenshuo Chang et al.
Journal of plant physiology, 167(14), 1152-1156 (2010-07-08)
Ethylene is an important plant gas hormone, and the amino acid Glu is emerging as a messenger molecule in plants. To evaluate the role of ethylene and Glu in seed germination and radicle growth under salt stress, effects of 1-aminocyclopropane-1-carboxylic
S Ali et al.
Journal of applied microbiology, 113(5), 1139-1144 (2012-07-24)
The ability of 1-aminocyclopropane-1-carboxylate (ACC) deaminase-containing plant growth-promoting bacterial (PGPB) endophytes Pseudomonas fluorescens YsS6 and Pseudomonas migulae 8R6, their ACC deaminase minus mutants and the rhizospheric plant growth-promoting bacterium Pseudomonas putida UW4 to delay the senescence of mini carnation cut
Yoram Kapulnik et al.
Journal of experimental botany, 62(8), 2915-2924 (2011-02-11)
Strigolactones (SLs) or derivatives thereof have been identified as phytohormones, and shown to act as long-distance shoot-branching inhibitors. In Arabidopsis roots, SLs have been suggested to have a positive effect on root-hair (RH) elongation, mediated via the MAX2 F-box. Two

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service