Skip to Content
Merck
All Photos(1)

Documents

05160

Sigma-Aldrich

β-Alanine

BioXtra, ≥99.0% (NT)

Synonym(s):

Beta-alanine, β-Ala, 3-Aminopropionic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2CH2CH2COOH
CAS Number:
Molecular Weight:
89.09
Beilstein:
906793
EC Number:
MDL number:
UNSPSC Code:
12352209
eCl@ss:
39030936
PubChem Substance ID:
NACRES:
NA.26

product line

BioXtra

Assay

≥99.0% (NT)

form

crystals

ign. residue

≤0.1% (as SO4)

loss

≤0.1% loss on drying, 20 °C (HV)

mp

~200 °C (dec.)
202 °C (dec.) (lit.)

anion traces

chloride (Cl-): ≤50 mg/kg
sulfate (SO42-): ≤50 mg/kg

cation traces

Ca: ≤10 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
K: ≤50 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Na: ≤50 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Zn: ≤5 mg/kg

application(s)

peptide synthesis

SMILES string

NCCC(O)=O

InChI

1S/C3H7NO2/c4-2-1-3(5)6/h1-2,4H2,(H,5,6)

InChI key

UCMIRNVEIXFBKS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • A new green approach to L-histidine and ß-alanine analysis in dietary supplements using rapid and simple contactless conductivity detection integrated with high-resolution glass-microchip electrophoresis.: This paper presents a novel method for analyzing ß-Alanine in dietary supplements, showcasing a green, efficient, and high-resolution technique (Pukleš et al., 2024).

Biochem/physiol Actions

β-Alanine, a β amino acid, is a component of pantothenic acid and the rate limiting amino acid in the biosynthesis of the histidinyl antioxidant dipeptides carnosine and anserine.
β-Alanine, a β−amino acid, is a component of pantothenic acid and the rate-limiting amino acid in the biosynthesis of the histidinyl antioxidant dipeptides carnosine and anserine. Endogenous β-amino acid that is a nonselective agonist at glycine receptors and a ligand for the G protein-coupled orphan receptor, TGR7 (MrgD). β-Alanine flux plays a cytoprotective role by supporting the osmotic stability of marine organisms, preimplantation mouse embryos and mammalian cells exposed to hypoxic stress.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jan Heyland et al.
Microbial biotechnology, 3(1), 74-83 (2011-01-25)
β-Peptides and their derivates are usually stable to proteolysis and have an increased half-life compared with α-peptides. Recently, β-aminopeptidases were described as a new enzyme class that enabled the enzymatic degradation and formation of β-peptides. As an alternative to the
Guilherme Giannini Artioli et al.
Medicine and science in sports and exercise, 42(6), 1162-1173 (2010-05-19)
In this narrative review, we present and discuss the current knowledge available on carnosine and beta-alanine metabolism as well as the effects of beta-alanine supplementation on exercise performance. Intramuscular acidosis has been attributed to be one of the main causes
Ramin Artang et al.
The American journal of cardiology, 112(12), 1973-1979 (2013-10-01)
Dabigatran has been associated with greater risk of myocardial infarction (MI) than warfarin. It is unknown whether the increased risk is unique to dabigatran, an adverse effect shared by other oral direct thrombin inhibitors (DTIs), or the result of a
Wilma Potze et al.
British journal of haematology, 163(5), 666-673 (2013-11-14)
There is increasing recognition that thrombotic complications may occur in patients with cirrhosis, and literature on antithrombotic treatment in these patients is rapidly emerging. Due to extensive haemostatic changes in patients with cirrhosis, careful monitoring of anticoagulant therapy may be
Torben Bjerregaard Larsen et al.
The American journal of medicine, 127(4), 329-336 (2013-12-24)
Dabigatran may provide less protection against myocardial infarction than vitamin K antagonists (VKAs) in patients with atrial fibrillation. This may be particularly evident among "switchers" to dabigatran from VKA, as a result of discontinuation effects. We identified in nationwide Danish

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service