Skip to Content
Merck

Skip To

01890

Adenosine

Synonym(s):

9-β-D-Ribofuranosyladenine, Adenine riboside, Adenine-9-β-D-ribofuranoside

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C10H13N5O4
CAS Number:
Molecular Weight:
267.24
PubChem Substance ID:
eCl@ss:
32160413
UNSPSC Code:
12352200
NACRES:
NA.77
EC Number:
200-389-9
MDL number:
Beilstein/REAXYS Number:
93029
Form:
powder
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

powder

Quality Segment

optical activity

[α]20/D −70±3°, c = 2% in 5% NaOH

ign. residue

≤0.1% (as SO4)

mp

234-236 °C (lit.)

cation traces

Ca: ≤10 mg/kg, Cd: ≤5 mg/kg, Co: ≤5 mg/kg, Cr: ≤5 mg/kg, Cu: ≤5 mg/kg, Fe: ≤5 mg/kg, Mg: ≤20 mg/kg, Mn: ≤5 mg/kg, Ni: ≤5 mg/kg, Pb: ≤5 mg/kg, Zn: ≤5 mg/kg

storage temp.

2-8°C

SMILES string

Nc1ncnc2n(cnc12)[C@@H]3O[C@H](CO)[C@@H](O)[C@H]3O

InChI

1S/C10H13N5O4/c11-8-5-9(13-2-12-8)15(3-14-5)10-7(18)6(17)4(1-16)19-10/h2-4,6-7,10,16-18H,1H2,(H2,11,12,13)/t4-,6-,7-,10-/m1/s1

InChI key

OIRDTQYFTABQOQ-KQYNXXCUSA-N

Gene Information

General description

Adenosine is a purine nucleoside base. It consists of adenine bound to a ribose sugar molecule via a glycosidic bond. Adenosine is found intracellularly and extracellularly in living cells. It acts as a precursor and as a metabolite of adenine nucleotides. In all living organisms, adenosine acts as an essential structural factor for adenosinetriphosphate (ATP), adenosine diphosphate (ADP), and cyclic adenosine monophosphate (cAMP), thereby playing an important role in energy transfer. Adenosine plays a crucial role in the central nervous system, endocrine system, inflammatory and immune response, and cardiovascular system. It is an important endogenous modulator of kidney function, lung function, pain, and sleep via the activation of adenosine receptors.

Application

Adenosine has been used as a standard to determine the adenosine content in whole heart samples from mice.

Biochem/physiol Actions

Endogenous neurotransmitter at adenosine receptors. Cardioprotective effects may relate to activation of A1 adenosine receptors. The antiplatelet and anti−inflammatory actions of adenosine appear to be mediated via the A2 adenosine receptor. In contrast, adenosine appears to be a pro-inflammatory mediator in asthma and chronic obstructive pulmonary disease (COPD).
Endogenous neurotransmitter. Cardioprotective effects may relate to activation of A1 adenosine receptors. The antiplatelet and anti−inflammatory actions of adenosine appear to be mediated via the A2 adenosine receptor. In contrast, adenosine appears to be a pro-inflammatory mediator in asthma and chronic obstructive pulmonary disease (COPD).

Other Notes

Substrate for the assay of adenosine deaminase

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
PHR11381012123A0230200
description

-

description

Pharmaceutical Secondary Standard; Certified Reference Material

description

United States Pharmacopeia (USP) Reference Standard

description

European Pharmacopoeia (EP) Reference Standard

Quality Level

200

Quality Level

300

Quality Level

-

Quality Level

-

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

-

storage temp.

2-8°C

form

powder

form

-

form

solid

form

-

Gene Information

human ... ADORA1(134), ADORA2A(135), ADORA2B(136), ADORA3(140)

Gene Information

human ... ADORA1(134), ADORA2A(135), ADORA2B(136), ADORA3(140)

Gene Information

human ... ADORA1(134), ADORA2A(135), ADORA2B(136), ADORA3(140)

Gene Information

human ... ADORA1(134), ADORA2A(135), ADORA2B(136), ADORA3(140)

cation traces

Ca: ≤10 mg/kg, Co: ≤5 mg/kg, Cu: ≤5 mg/kg, Mg: ≤20 mg/kg, Ni: ≤5 mg/kg, Zn: ≤5 mg/kg, Cd: ≤5 mg/kg, Fe: ≤5 mg/kg, Pb: ≤5 mg/kg, Cr: ≤5 mg/kg, Mn: ≤5 mg/kg

cation traces

-

cation traces

-

cation traces

-

optical activity

[α]20/D −70±3°, c = 2% in 5% NaOH

optical activity

-

optical activity

-

optical activity

-


Still not finding the right product?

Explore all of our products under Adenosine


Storage Class

11 - Combustible Solids

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters