Skip to Content
Merck
All Photos(2)

Key Documents

H13303

Sigma-Aldrich

1-Hexanol

reagent grade, 98%

Synonym(s):

Hexyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)5OH
CAS Number:
Molecular Weight:
102.17
Beilstein:
969167
EC Number:
MDL number:
UNSPSC Code:
12352104
PubChem Substance ID:
NACRES:
NA.21
grade:
reagent grade
Assay:
98%
bp:
156-157 °C (lit.)
vapor pressure:
1 mmHg ( 25.6 °C)

grade

reagent grade

Quality Level

vapor density

4.5 (vs air)

vapor pressure

1 mmHg ( 25.6 °C)

Assay

98%

form

liquid

autoignition temp.

559 °F

expl. lim.

0.34-6.3 %

refractive index

n20/D 1.418 (lit.)

bp

156-157 °C (lit.)

mp

−52 °C (lit.)

density

0.814 g/mL at 25 °C (lit.)

SMILES string

CCCCCCO

InChI

1S/C6H14O/c1-2-3-4-5-6-7/h7H,2-6H2,1H3

InChI key

ZSIAUFGUXNUGDI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1-Hexanol is a linear primary alcohol, made up of six carbon atoms and is produced by the oligomerization of ethylene. There is a wide range of uses for 1-hexanol, including plasticizers, antiseptics, fragrances, pharmaceuticals, textiles, and leather finishing agents.

Application

1-Hexanol is used:
  • As an oil phase in the preparation of manganese zinc ferrite nanoparticles by precipitation in reverse microemulsion system.
  • As a solvent in the separation of carboxylic acids and tetrahydrofurfuryl alcohol from water.
  • As a solvent in the synthesis of ZnO (zinc oxide) quantum particles from zinc acetate by precipitation method.

Features and Benefits

1-Hexanol or Hexyl alcohol exhibits high reactivity at a lower temperature due to the alkane-like reaction pathways that begin at a secondary carbon site.

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

140.0 °F - closed cup

Flash Point(C)

60 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Erik Jue et al.
Scientific reports, 10(1), 1940-1940 (2020-02-08)
The success of fundamental and applied nucleic acid (NA) research depends on NA purity, but obtaining pure NAs from raw, unprocessed samples is challenging. Purification using solid-phase NA extractions utilizes sequential additions of lysis and wash buffers followed by elution.
Maria Eugenia Villar et al.
Cell reports, 30(8), 2603-2613 (2020-02-27)
Research on honeybee memory has led to a widely accepted model in which a single pairing of an odor stimulus with sucrose induces memories that are independent of protein synthesis but is unable to form protein-synthesis-dependent long-term memory (LTM). The
Justin T Mohr et al.
Journal of medicinal chemistry, 48(12), 4172-4176 (2005-06-10)
The polyhydroxyalkanes 1,6,11,16-hexadecanetetraol (1) and 2,7,12,17-octadecanetetraol (2) were synthesized utilizing the thiophene ring as a scaffold to affix the hydroxyalkyl chains by lithiation of the acidic alpha-hydrogens and subsequent desulfurization. Both compounds exhibited significant anesthetic potency, individually and in additivity
Annie Handler et al.
Cell, 178(1), 60-75 (2019-06-25)
Animals rely on the relative timing of events in their environment to form and update predictive associations, but the molecular and circuit mechanisms for this temporal sensitivity remain incompletely understood. Here, we show that olfactory associations in Drosophila can be
Zhisong Lu et al.
Nanotechnology, 22(15), 155604-155604 (2011-03-11)
With advances of quantum dots (QDs) in bioimaging applications, various materials have been used to coat QDs to reduce their nanotoxicity; however, the coating could introduce new toxic sources and quench the fluorescence in bioimaging applications. In this work, ZrO₂

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service