Skip to Content
Merck
All Photos(8)

Key Documents

746363

Sigma-Aldrich

Sodium sulfate

anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Na2SO4
CAS Number:
Molecular Weight:
142.04
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21

grade

ACS reagent
anhydrous

Quality Level

Agency

suitable for EPA 1613

product line

Redi-Dri

Assay

≥99%

form

powder

quality

free-flowing

impurities

≤0.01% insolubles
≤5 ppm N compounds

loss

≤0.5% loss on ignition

pH

5.2-9.2 (25 °C, 5%)

mp

884 °C (lit.)

density

2.68 g/mL at 25 °C (lit.)

anion traces

chloride (Cl-): ≤0.001%
phosphate (PO43-): ≤0.001%

cation traces

Ca: ≤0.002%
Fe: ≤0.001%
K: ≤0.002%
Mg: ≤0.001%
heavy metals: ≤5 ppm (by ICP-OES)

SMILES string

[Na+].[Na+].[O-]S([O-])(=O)=O

InChI

1S/2Na.H2O4S/c;;1-5(2,3)4/h;;(H2,1,2,3,4)/q2*+1;/p-2

InChI key

PMZURENOXWZQFD-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Sodium sulfate can be used as a dehydrating agent for the synthesis of 1,3-diaryl-2-propen-1-one oxime via the condensation of 1,3-diaryl-2-propen-1-one with hydroxylamine hydrochloride. It can also be used as a drying agent in organic synthesis for the removal of traces of water from organic extracts. It is used for drying almost all compounds including fatty acids, aldehydes, ketones, and alkyl and aryl halogenides.

Features and Benefits

  • Increased efficiency – Eliminates time and effort of loosening hardened powders     
  • Safety – Eliminates tools of force used to break up clumps
  • Ecology – Reduces waste as it does not stick to the packaging
  • Economy – Faster preparation and solvation increase productivity and reduce costs      
  • Assured quality – Excellent, expert-tested quality with no anti-caking agents      
  • Flexible volumes – Available from research amounts to scale-up quantities

Legal Information

Redi-Dri is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Teppei Adachi et al.
Inflammatory bowel diseases, 21(1), 31-39 (2014-12-02)
Expression of heat shock protein A4 (HSPA4, also called Apg-2), a member of the HSP110 family, is induced by several forms of stress. The physiological and pathological functions of HSPA4 in the intestine remain to be elucidated. We assessed HSPA4
Christoph Zürcher et al.
Journal of virology, 88(13), 7235-7243 (2014-04-18)
The RNase activity of the envelope glycoprotein E(rns) of the pestivirus bovine viral diarrhea virus (BVDV) is required to block type I interferon (IFN) synthesis induced by single-stranded RNA (ssRNA) and double-stranded RNA (dsRNA) in bovine cells. Due to the
Rebecca L Scalzo et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 28(6), 2705-2714 (2014-03-07)
Improved endurance exercise performance in adult humans after sprint interval training (SIT) has been attributed to mitochondrial biogenesis. However, muscle protein synthesis (MPS) and mitochondrial biogenesis during SIT have not been measured, nor have sex-specific differences. We hypothesized that males
Jiao Zhao et al.
Angewandte Chemie (International ed. in English), 53(44), 11808-11812 (2014-09-12)
Porous films of p-type CuInS2, prepared by sulfurization of electrodeposited metals, are surface-modified with thin layers of CdS and TiO2. This specific porous electrode evolved H2 from photoelectrochemical water reduction under simulated sunlight. Modification with thin n-type CdS and TiO2
Erik C Dreaden et al.
ACS nano, 8(8), 8374-8382 (2014-08-08)
Active targeting of nanoscale drug carriers can improve tumor-specific delivery; however, cellular heterogeneity both within and among tumor sites is a fundamental barrier to their success. Here, we describe a tumor microenvironment-responsive layer-by-layer (LbL) polymer drug carrier that actively targets

Articles

Redi-Dri™ prevents hygroscopic powders, such as inorganic salts, from absorbing moisture and forming clumps, leaving the salts free-flowing every time.

Redi-Dri™ prevents hygroscopic powders, such as inorganic salts, from absorbing moisture and forming clumps, leaving the salts free-flowing every time.

Redi-Dri™ prevents hygroscopic powders, such as inorganic salts, from absorbing moisture and forming clumps, leaving the salts free-flowing every time.

Redi-Dri™ prevents hygroscopic powders, such as inorganic salts, from absorbing moisture and forming clumps, leaving the salts free-flowing every time.

Related Content

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service