Skip to Content
Merck
All Photos(1)

Key Documents

676918

Sigma-Aldrich

Carbon disulfide

ACS reagent, ≥99.9%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CS2
CAS Number:
Molecular Weight:
76.14
Beilstein:
1098293
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21
grade:
ACS reagent
bp:
46 °C (lit.)
vapor pressure:
5.83 psi
Pricing and availability is not currently available.

grade

ACS reagent

vapor density

2.67 (vs air)

vapor pressure

5.83 psi

Assay

≥99.9%

form

liquid

autoignition temp.

212 °F

expl. lim.

50 %

dilution

(for analytical testing)

impurities

H2S, passes test (lim. ~1.5 ppm)
SO2, passes test (lim. ~2.5 ppm)
≤0.05% water

evapn. residue

≤0.002%

color

APHA: ≤10

refractive index

n20/D 1.627 (lit.)

bp

46 °C (lit.)

mp

−112-−111 °C (lit.)

density

1.266 g/mL at 25 °C (lit.)

SMILES string

S=C=S

InChI

1S/CS2/c2-1-3

InChI key

QGJOPFRUJISHPQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Carbon disulfide is a widely used solvent in the production of rayon fibers, carbon tetrachloride, pesticides, vulcanizers, cellophane, adhesives, and other chemicals. It is also used as a starting material in the synthesis of various sulfur-containing derivatives.

Application

Carbon disulfide can be used as a solvent:
  • To synthesize hydroxynaphthyl ketones via Friedel-Crafts acylation and demethylation.
  • In the regioselective bromination of binaphthols.

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Eye Irrit. 2 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1

Target Organs

Peripheral nervous system,Central nervous system,Cardio-vascular system,Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

-22.0 °F - closed cup

Flash Point(C)

-30 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Mark B van Eldijk et al.
Chemical communications (Cambridge, England), 49(71), 7770-7772 (2013-06-19)
CS2 hydrolase, a zinc-dependent enzyme that converts carbon disulfide to carbon dioxide and hydrogen sulfide, exists as a mixture of octameric ring and hexadecameric catenane forms in solution. A combination of size exclusion chromatography, multi-angle laser light scattering, and mass
Continuous direct spinning of fibers of single-walled carbon nanotubes with metallic chirality.
Rajyashree M Sundaram et al.
Advanced materials (Deerfield Beach, Fla.), 23(43), 5064-5068 (2011-10-11)
Huan Liu et al.
Journal of hazardous materials, 235-236, 298-306 (2012-08-21)
Chemical conditioners are often used to enhance sewage sludge dewaterability through altering sludge properties and flocs structure, both affect odorous compounds emissions not only during sludge conditioning but also in subsequent sludge disposal. This study was to investigate emission characteristics
R W Wood
Neurobehavioral toxicology and teratology, 3(4), 397-405 (1981-01-01)
Carbon disulfide is an exceptionally potent neurotoxicant. In industrial settings its effects are pernicious, producing symptoms and behavioral alterations in the absence of frank poisoning; affective disorders are common, and exposure is associated with an elevated incidence of suicide. With
Toxicology of carbon disulfide; a review.
R W Coppock et al.
Veterinary and human toxicology, 23(5), 331-336 (1981-10-01)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service