Skip to Content
Merck
All Photos(1)

Key Documents

63547

Sigma-Aldrich

Manganese(II) nitrate tetrahydrate

purum p.a., ≥97.0% (KT)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Mn(NO3)2 · 4H2O
CAS Number:
Molecular Weight:
251.01
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21
Assay:
≥97.0% (KT)
form:
powder, crystals or chunks

grade

purum p.a.

Quality Level

Assay

≥97.0% (KT)

form

powder, crystals or chunks

anion traces

chloride (Cl-): ≤50 mg/kg
sulfate (SO42-): ≤50 mg/kg

cation traces

Ca: ≤50 mg/kg
Cd: ≤50 mg/kg
Co: ≤50 mg/kg
Cu: ≤50 mg/kg
Fe: ≤50 mg/kg
K: ≤100 mg/kg
Na: ≤100 mg/kg
Ni: ≤50 mg/kg
Pb: ≤50 mg/kg
Zn: ≤50 mg/kg

SMILES string

O.O.O.O.[Mn++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

InChI

1S/Mn.2NO3.4H2O/c;2*2-1(3)4;;;;/h;;;4*1H2/q+2;2*-1;;;;

InChI key

ALIMWUQMDCBYFM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Manganese(II) nitrate tetrahydrate is a hydrated manganese salt.

Application

Manganese(II) nitrate tetrahydrate may be employed as starting reagent in the preparation of:
  • [Ni1/3Co1/3Mn1/3]O2 powders
  • [Mn(2-Bzpy)(N3)2]n (2-Benzy = 2-benzoylpyridine)
  • [Mn(3,5-lutidine)2(N3)2]n

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Ox. Sol. 3 - Skin Corr. 1C - STOT RE 2 Inhalation

Target Organs

Brain

Supplementary Hazards

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 2


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis, structural characterization, magnetic behavior, and single crystal EPR spectra of three new one-dimensional manganese azido systems with FM, alternating FM-AF, and AF coupling.
Abu-Youssef MAM, et al.
Inorganic Chemistry, 38(25), 5716-5723 (1999)
Nils Lau Nyborg Broge et al.
Nanoscale, 12(15), 8511-8518 (2020-04-04)
Solvothermal technology shows great promise in "green" materials synthesis, processing, and recycling. The outcome of a specific solvothermal reaction depends strongly on the solvent properties, and the versatility of solvothermal synthesis hinges on the very large changes in solvent properties
Synthesis and structural characterization of layered Li[Ni1/3Co1/3Mn1/3]O2 cathode materials by ultrasonic spray pyrolysis method.
Park SH, et al.
Electrochimica Acta, 49(4), 557-563 (2004)
Longwen Chen et al.
Journal of environmental sciences (China), 74, 58-70 (2018-10-21)
Processes based on non-thermal plasma (NTP) for indoor air treatment inevitably lead to the formation of toxic by-products such as ozone (O3) and nitrogen oxides (NOx). Adding a step of heterogeneous catalysis in series with NTP could allow for the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service