Skip to Content
Merck
All Photos(1)

Key Documents

456438

Sigma-Aldrich

1,2,3,4-Tetrahydronaphthalene

reagent grade, ≥97%

Synonym(s):

Tetralin solvent

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H12
CAS Number:
Molecular Weight:
132.20
Beilstein:
1446407
EC Number:
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
NACRES:
NA.21
grade:
reagent grade
bp:
207 °C (lit.)
vapor pressure:
0.18 mmHg ( 20 °C)

grade

reagent grade

Quality Level

vapor density

4.55 (vs air)

vapor pressure

0.18 mmHg ( 20 °C)

Assay

≥97%

form

liquid

autoignition temp.

723 °F

expl. lim.

0.8 %, 100 °F
5 %, 150 °F

refractive index

n20/D 1.541 (lit.)

bp

207 °C (lit.)

mp

−35 °C (lit.)

density

0.973 g/mL at 25 °C (lit.)

SMILES string

C1CCc2ccccc2C1

InChI

1S/C10H12/c1-2-6-10-8-4-3-7-9(10)5-1/h1-2,5-6H,3-4,7-8H2

InChI key

CXWXQJXEFPUFDZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Tetralin is a bicyclic molecule composed of an aromatic moiety and an alicyclic moiety that share two carbon atoms in common. It is used as a hydrogen-donor solvent in coal liquefaction. Tetralin is also used to synthesize agricultural chemicals. Additionally, it is used as a degreaser and solvent for fats, resins, and waxes and as a substitute for turpentine in paints, lacquers, and shoe polishes.

Application

1,2,3,4-Tetrahydronaphthalene is used as a:
  • Hydrogen-donor solvent in the decarboxylation of oleic acid to produce diesel fuel hydrocarbons.
  • Hydrogen source for the hydrogenation of lignin-derived phenolic compounds.

Legal Information

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2

Supplementary Hazards

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

159.8 °F - closed cup

Flash Point(C)

71 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fullerene production in sooting flames from 1,2,3,4-tetrahydronaphthalene.
Alford JM, et al.
Carbon, 46(12), 1623-1625 (2008)
Activated carbon supported molybdenum carbides as cheap and highly efficient catalyst in the selective hydrogenation of naphthalene to tetralin.
Pang M, et al.
Green Chemistry, 14(5), 1272-1276 (2012)
Experimental and kinetic modeling study of tetralin pyrolysis at low pressure.
Li Y, et al.
Proceedings of the Combustion Institute, 34(1), 1739-1748 (2013)
The Low-Temperature Thermodynamic Properties of Naphthalene, 1-Methylnaphthalene, 2-Methylnaphthalene, 1,2,3,4-Tetrahydronaphthalene, trans-Decahydronaphthalene and cis-Decahydronaphthalene.
McCullough JP, et al.
The Journal of Physical Chemistry, 61(8), 1105-1116 (1957)
Hydrogen production from tetralin over microwave-accelerated Pt-supported activated carbon.
Suttisawat Y, et al.
International Journal of Hydrogen Energy, 35(12), 6179-6183 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service