Skip to Content
Merck
All Photos(2)

Documents

435597

Sigma-Aldrich

Sulfuric acid, fuming

reagent grade, 20%, as free SO3

Synonym(s):

Oleum

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2SO4 · (SO3)x
CAS Number:
Molecular Weight:
98.08
Beilstein:
2037554
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.21

grade

reagent grade

Quality Level

vapor density

3 (vs air)

vapor pressure

2 mmHg ( 25 °C)

form

liquid

concentration

20% (free SO3)

density

1.925 g/mL at 25 °C (lit.)

SMILES string

O=S(=O)=O.OS(O)(=O)=O

InChI

1S/H2O4S.O3S/c1-5(2,3)4;1-4(2)3/h(H2,1,2,3,4);

InChI key

HIFJUMGIHIZEPX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Fuming sulfuric acid is an inorganic acid, that can be produced by the addition of sulfur trioxide to concentrated sulfuric acid. It is commonly used as a sulfonating reagent in sulfonation reactions. When fuming sulfuric acid is employed as reaction medium, bromination takes place only in the pyridine nucleus of isomeric methylpyridines and 2,6-dimethylpyridine. It can also oxidize fullerene C60 to form fullerene polymers.

Application

Fuming sulfuric acid may be used as a sulfonating agent for the synthesis of:
  • sulfonated poly(1,4-phenylene sulfide)
  • water-soluble polyphosphazene polyelectrolyte
  • sulfated graphene

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Inhalation - Skin Corr. 1A - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Daley RF and Daley SJ.
Organic Chemistry, Part 3 of 3, 801-801 null
Bromination of methylpyridines in fuming sulfuric acid.
Van der Does L and Hertog HJD.
Rec. Trav. Chim., 84(7), 951-964 (1965)
Products of reaction of fullerene C60 with fuming sulfuric acid studied by IR and ESR spectroscopy.
Garkusha OG, et al.
Russian Chemical Bulletin, 51(4), 628-631 (2002)
Sulfated graphene as an efficient solid catalyst for acid-catalyzed liquid reactions.
Liu F, et al.
Journal of Materials Chemistry, 22(12), 5495-5502 (2012)
Proton conducting composite membranes comprising sulfonated Poly(1,4-phenylene sulfide) and zeolite for fuel cell.
Choi JM, et al.
Ionics, 16(5), 403-408 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service