Skip to Content
Merck
All Photos(3)

Key Documents

32319

Sigma-Aldrich

Sodium acetate

puriss. p.a., ACS reagent, reag. Ph. Eur., anhydrous

Synonym(s):

Acetic acid sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COONa
CAS Number:
Molecular Weight:
82.03
Beilstein:
3595639
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21

grade

ACS reagent
puriss. p.a.

Agency

reag. Ph. Eur.

Assay

≥99.0% (perchloric acid titration)

form

solid

autoignition temp.

1112 °F

impurities

≤0.01% in water soluble matter

loss

≤1% loss on drying, 120 °C

pH

7.5-9.2 (20 °C, 5%)

pKa 

4.76 (acetic acid)

mp

>300 °C (dec.) (lit.)

anion traces

chloride (Cl-): ≤0.002%
phosphate (PO43-): ≤0.001%
sulfate (SO42-): ≤0.003%

cation traces

Al: ≤10 mg/kg
Ca: ≤0.005%
Cd: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤0.001%
K: ≤200 mg/kg
Mg: ≤0.002%
Pb: ≤5 mg/kg
Zn: ≤5 mg/kg
heavy metals: ≤0.001% (by ICP-OES)

suitability

in accordance for appearance of solution

SMILES string

[Na+].CC([O-])=O

InChI

1S/C2H4O2.Na/c1-2(3)4;/h1H3,(H,3,4);/q;+1/p-1

InChI key

VMHLLURERBWHNL-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Sodium acetate can be used as a catalyst to synthesize:
  • 1,2-Dihydropyridines by reacting 3-formylchromones, N-nucleophiles, and active methylene molecules.
  • 2(3H)-Furanone derivatives by using cinnamaldehyde and 5-aryl-2(3H) furanone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Facile Route to Synthesize 1, 2-Dihydropyridine and 1, 6 Pyrimidinone Analogues from 4-Oxo-4H-Chromene-3-Carbaldehyde
Bari A
Journal of Heterocyclic Chemistry, 53, 461-466 (2016)
Synthesis, Characterization and Photophysical Properties of New 2 (3H) Furanone Derivatives
Soliman M, et al.
Journal of Molecular Structure, 134543-134543 (2022)
Carol L Ladner-Keay et al.
PloS one, 9(6), e98753-e98753 (2014-06-04)
The formation of β-sheet rich prion oligomers and fibrils from native prion protein (PrP) is thought to be a key step in the development of prion diseases. Many methods are available to convert recombinant prion protein into β-sheet rich fibrils
Mari Endo et al.
Journal of gastroenterology, 49(6), 1026-1039 (2013-07-13)
Daikenchuto (DKT), a gastrointestinal prokinetic Japanese herbal medicine, is prescribed for patients with postoperative ileus (POI) and adhesive bowel obstruction following abdominal surgery. Several mechanisms for the amelioration of POI by DKT have been suggested; however, it has remained unclear
Andrew D Shaw et al.
Annals of surgery, 255(5), 821-829 (2012-04-04)
To assess the association of 0.9% saline use versus a calcium-free physiologically balanced crystalloid solution with major morbidity and clinical resource use after abdominal surgery. 0.9% saline, which results in a hyperchloremic acidosis after infusion, is frequently used to replace

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service