Skip to Content
Merck
All Photos(1)

Key Documents

30755

Sigma-Aldrich

Perchloric acid

puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., 70.0-72.0%

Synonym(s):

PCA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HClO4
CAS Number:
Molecular Weight:
100.46
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:

grade

ACS reagent
puriss. p.a.

Agency

USP/NF
reag. ISO
reag. Ph. Eur.

Assay

70.0-72.0%

form

liquid

impurities

≤0.00005% free chlorine (Cl)
≤0.0001% heavy metals (as Pb)
≤0.001% total nitrogen (N)

ign. residue

≤0.003% (as SO4)

density

1.670 g/cm3

anion traces

chlorate (ClO3-): ≤10 mg/kg
chloride (Cl-): ≤3 mg/kg
phosphate, silicate (as SiO2): ≤5 mg/kg
sulfate (SO42-): ≤10 mg/kg

cation traces

Ag: ≤0.1 mg/kg
Al: ≤0.05 mg/kg
As: ≤0.05 mg/kg
Ba: ≤0.02 mg/kg
Be: ≤0.02 mg/kg
Bi: ≤0.1 mg/kg
Ca: ≤0.5 mg/kg
Cd: ≤0.05 mg/kg
Co: ≤0.05 mg/kg
Cu: ≤0.1 mg/kg
Fe: ≤1 mg/kg
Ge: ≤0.05 mg/kg
K: ≤0.1 mg/kg
Li: ≤0.02 mg/kg
Mg: ≤0.5 mg/kg
Mn: ≤0.02 mg/kg
Mo: ≤0.05 mg/kg
Ni: ≤0.1 mg/kg
Pb: ≤0.05 mg/kg
Sr: ≤0.02 mg/kg
Ti: ≤0.1 mg/kg
Tl: ≤0.05 mg/kg
V: ≤0.05 mg/kg
Zn: ≤0.1 mg/kg
Zr: ≤0.1 mg/kg

SMILES string

OCl(=O)(=O)=O

InChI

1S/ClHO4/c2-1(3,4)5/h(H,2,3,4,5)

InChI key

VLTRZXGMWDSKGL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Perchloric acid (HClO4) is a strong mineral acid. It has powerful oxidizing and dehydrating properties when it is hot and concentrated. It is commercially available in a concentration of 70% (by weight) in water. Aqueous solution of HClO4 is regarded as a super acid (stronger than sulfuric and nitric acids), but it is not necessarily a powerful oxidizer.

Application

Perchloric acid can be used:      
  • In the synthesis of graphite oxide via electrochemical oxidation of natural graphite.      
  • As a reaction medium in oxidations reactions.     
  • As a dopant in the preparation of perchloric acid-functionalized polyaniline (HClO4/PANI) catalyst for the synthesis of 2-substituted benzothiazoles by condensation reaction between o-aminothiophenol with aldehydes.

It can also be used in the following:      
  • Silica-supported perchloric acid (HClO4-SiO2) is used as a catalyst to synthesize amidoalkyl naphthols by the condensation reaction of 2-naphthol, aromatic aldehydes, and acetonitrile.      
  • HClO4-SiO2 is used in the Friedlander quinoline synthesis by the condensation reaction of 2-aminoaryl ketones and carbonyl compounds.      
  • Alumina-supported perchloric acid (Al2O3-HClO4) is used as a catalyst in the synthesis of α-(α-amidobenzyl)-β-naphthols.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Met. Corr. 1 - Ox. Liq. 1 - Skin Corr. 1A - STOT RE 2

Target Organs

Thyroid

Storage Class Code

5.1A - Strongly oxidizing hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Sulfur in plant materials by digestion with nitric and perchloric acid.
Blanchar RW, et al.
Soil Science Society of America Journal. Soil Science Society of America, 29(1), 71-72 (1965)
Eagleson M.
Concise Encyclopedia Chemistry, 790-790 (1994)
Taro Takami et al.
Cancer research, 67(20), 9844-9851 (2007-10-19)
Hepatocyte growth factor (HGF) has been reported to have both positive and negative effects on carcinogenesis. Here, we show that the loss of c-Met signaling in hepatocytes enhanced rather than suppressed the early stages of chemical hepatocarcinogenesis. c-Met conditional knockout
Martin G Liebensteiner et al.
Science (New York, N.Y.), 340(6128), 85-87 (2013-04-06)
Perchlorate and chlorate anions [(per)chlorate] exist in the environment from natural and anthropogenic sources, where they can serve as electron acceptors for bacteria. We performed growth experiments combined with genomic and proteomic analyses of the hyperthermophile Archaeoglobus fulgidus that show
Dipanwita Das et al.
Journal of the American Chemical Society, 135(10), 4018-4026 (2013-02-28)
Catalytic four-electron reduction of O2 by ferrocene (Fc) and 1,1'-dimethylferrocene (Me2Fc) occurs efficiently with a dinuclear copper(II) complex [Cu(II)2(XYLO)(OH)](2+) (1), where XYLO is a m-xylene-linked bis[(2-(2-pyridyl)ethyl)amine] dinucleating ligand with copper-bridging phenolate moiety], in the presence of perchloric acid (HClO4) in

Articles

Using an Ascentis® Express F5 HPLC column, this study separates 11 β-agonists in pork by LC-MS/MS in under 8 minutes with excellent recovery and precision.

Using an Ascentis® Express F5 HPLC column, this study separates 11 β-agonists in pork by LC-MS/MS in under 8 minutes with excellent recovery and precision.

Using an Ascentis® Express F5 HPLC column, this study separates 11 β-agonists in pork by LC-MS/MS in under 8 minutes with excellent recovery and precision.

Using an Ascentis® Express F5 HPLC column, this study separates 11 β-agonists in pork by LC-MS/MS in under 8 minutes with excellent recovery and precision.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service