Skip to Content
Merck
All Photos(3)

Key Documents

293261

Sigma-Aldrich

4-Methyl-2-pentanone

suitable for HPLC, ≥99.5%

Synonym(s):

Isobutyl methyl ketone, Isopropylacetone, MIBK, Methyl isobutyl ketone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHCH2COCH3
CAS Number:
Molecular Weight:
100.16
Beilstein:
605399
EC Number:
MDL number:
UNSPSC Code:
12190000
PubChem Substance ID:
NACRES:
NA.21

vapor density

3.5 (vs air)

vapor pressure

15.7 mmHg

Assay

≥99.5%

form

liquid

autoignition temp.

840 °F

expl. lim.

1.2-8 %, 93 °F

technique(s)

HPLC: suitable

impurities

<0.050% water

evapn. residue

<0.001%

refractive index

n20/D 1.395 (lit.)

bp

117-118 °C

mp

−80 °C (lit.)

density

0.801 g/mL at 25 °C (lit.)

λ

H2O reference

UV absorption

λ: 335 nm Amax: 1.00
λ: 340 nm Amax: 0.50
λ: 360 nm Amax: 0.15
λ: 380 nm Amax: 0.02
λ: 400 nm Amax: 0.01

application(s)

food and beverages

SMILES string

CC(C)CC(C)=O

InChI

1S/C6H12O/c1-5(2)4-6(3)7/h5H,4H2,1-3H3

InChI key

NTIZESTWPVYFNL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This solvent is glass distilled, submicron filtered and subjected to rigorous specification testing to provide lot-to-lot consistency.

Application

Designed for use with HPLC instrumentation and organic synthesis applications.
Featured as a solvent in the extraction of cellulose-derived levulinic acid from municipal wastewater treatment plant (WWTP) sludge prior to HPLC analysis.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Carc. 2 Inhalation - Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

57.2 °F - closed cup

Flash Point(C)

14 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mini Mol Menamparambath et al.
Nanoscale, 6(15), 8844-8851 (2014-06-24)
A difference in work function plays a key role in charge transfer between two materials. Inorganic electrides provide a unique opportunity for electron transfer since interstitial anionic electrons result in a very low work function of 2.4-2.6 eV. Here we
Catalytic behaviors of silica-supported starch-polysulfosiloxane-Pt complexes in asymmetric hydrogenation of 4-methyl-2-pentanone.
Huang K, et al.
Reactive functional Polymers, 50(3), 199-203 (2003)
Excess volumes and viscosities of binary systems containing 4-methyl-2-pentanone.
Fermeglia M, et al.
Journal of Chemical and Engineering Data, 35(3), 260-265 (1990)
H J Shim et al.
Journal of chromatography. B, Biomedical applications, 656(2), 407-414 (1994-06-17)
A high-performance liquid chromatographic method was developed for the simultaneous determination of a new anthracycline, DA-125 (I), and its metabolites (M1, M2, M3, and M4) in rat plasma and urine using fluorescein as an internal standard. Compound I, a prodrug
Ginette Truchon et al.
Journal of occupational and environmental hygiene, 3(3), 137-143 (2006-02-09)
Compartmental and physiologically based toxicokinetic modeling coupled with Monte Carlo simulation were used to quantify the impact of biological variability (physiological, biochemical, and anatomic parameters) on the values of a series of bio-indicators of metal and organic industrial chemical exposures.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service