Skip to Content
Merck
All Photos(1)

Key Documents

249882

Sigma-Aldrich

Aluminum chloride solution

1.0 M in nitrobenzene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
AlCl3
CAS Number:
Molecular Weight:
133.34
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.55
grade:
for analytical purposes
form:
liquid
Pricing and availability is not currently available.

grade

for analytical purposes

form

liquid

reaction suitability

reagent type: catalyst
core: aluminum

concentration

0.95-1.10 M (EDTA titration)
1.0 M in nitrobenzene

density

1.249 g/mL at 25 °C

SMILES string

Cl[Al](Cl)Cl

InChI

1S/Al.3ClH/h;3*1H/q+3;;;/p-3

InChI key

VSCWAEJMTAWNJL-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 3 - Carc. 2 - Eye Dam. 1 - Repr. 1B - Skin Corr. 1B - STOT RE 1 Inhalation

Target Organs

Blood

Supplementary Hazards

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

190.4 °F - closed cup

Flash Point(C)

88 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Amination of< i> exo</i>-2-chloronorbornane and norbornane with trichloramine-aluminum chloride.
Kovacic P, et al.
Tetrahedron, 26(2), 529-538 (1970)
Zhiliang Huang et al.
Organic & biomolecular chemistry, 11(11), 1810-1814 (2013-02-08)
AlCl(3) promoted Friedel-Crafts acylation between 4-tert-butylbenzoyl chloride and mesitylene was investigated. The donor-acceptor complex was observed as the major species. Kinetic investigation demonstrated that the reaction was first-order on the donor-acceptor complex and zero-order on ArH, suggesting that the donor-acceptor
Acceleration of the Diels-Alder reaction by aluminum chloride.
Yates P and Eaton P.
Journal of the American Chemical Society, 82(16), 4426-4437 (1960)
Diagnosis and management of hyperhidrosis.
R A Benson et al.
BMJ (Clinical research ed.), 347, f6800-f6800 (2013-11-28)
Hai-hua Zhao et al.
Behavioural brain research, 241, 228-234 (2012-12-12)
Environmental agent aluminum, a well-known neurotoxin, has been proposed to play a role in the development of Alzheimer's disease (AD), and produced clinical and pathological features which were strikingly similar to those seen in AD brain, such as neurofibrillary tangles.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service