Skip to Content
Merck
All Photos(3)

Key Documents

236470

Sigma-Aldrich

Chromium(VI) oxide

ACS reagent, ≥98.0%

Synonym(s):

Chromic anhydride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CrO3
CAS Number:
Molecular Weight:
99.99
EC Number:
MDL number:
UNSPSC Code:
12352303
PubChem Substance ID:
NACRES:
NB.24
Assay:
≥98.0%
grade:
ACS reagent
form:
powder or crystals

grade

ACS reagent

Quality Level

Assay

≥98.0%

form

powder or crystals

reaction suitability

reagent type: oxidant

impurities

≤0.01% insolubles

pH

<1 (20 °C, 100 g/L)

mp

196 °C (dec.) (lit.)

anion traces

chloride (Cl-): ≤0.005%
nitrate (NO3-): ≤0.05%
sulfate (SO42-): ≤0.005%

cation traces

Fe, Al, Ba: ≤0.03%
Na: ≤0.2%

SMILES string

O=[Cr](=O)=O

InChI

1S/Cr.3O

InChI key

WGLPBDUCMAPZCE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chromium(VI) oxide is an oxidizing reagent for the oxidation of carbon-hydrogen bonds to alcohols, alkylaromatics to ketones and carboxylic acids, alkenes to α, β-unsaturated ketones, arenes to quinones, and alcohols to aldehydes, ketones, acids, and keto acids.

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1A - Eye Dam. 1 - Muta. 1B - Ox. Sol. 1 - Repr. 2 - Resp. Sens. 1 - Skin Corr. 1A - Skin Sens. 1 - STOT RE 1 Inhalation - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

5.1A - Strongly oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chromium (VI) Oxide
Freeman F, et al.
e-EROS Encyclopedia of Reagents for Organic Synthesis (2001)
Chromium (VI) Oxide.
Freeman F, et al.
e-EROS Encyclopedia of Reagents for Organic Synthesis. (2008)
Chromium (VI) oxide-catalyzed oxidation of arenes with periodic acid.
Yamazaki S.
Tetrahedron Letters, 42(19), 3355-3357 (2001)
L Vayssieres et al.
Journal of nanoscience and nanotechnology, 1(4), 385-388 (2003-08-14)
We are reporting here on the inexpensive fabrication and optical properties of an iron(III) oxide-chromium(III) oxide nanocomposite thin film of corundum crystal structure. Its novel and unique-designed architecture consists of uniformed, well-defined and oriented nanorods of Hematite (alpha-Fe2O3) of 50
M C García-Rodríguez et al.
Mutation research, 496(1-2), 145-151 (2001-09-12)
The effect of chlorophyllin on micronucleated polychromatic erythrocytes (MN-PCE) induction by chromium trioxide (CrO(3)) exposure in peripheral blood of mice was studied. Animals were treated with a single intraperitoneal dose of chlorophyllin (CHL) (20mg/kg), CrO(3) (20mg/kg), and CHL (20mg/kg) 4h

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service