Skip to Content
Merck
All Photos(3)

Key Documents

222003

Sigma-Aldrich

Sodium phosphate tribasic dodecahydrate

ACS reagent, ≥98%

Synonym(s):

TSP, Trisodium phosphate (tert) dodecahydrate, Trisodium phosphate dodecahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Na3PO4 · 12H2O
CAS Number:
Molecular Weight:
380.12
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21
Assay:
≥98%
grade:
ACS reagent
form:
crystals or chunks

grade

ACS reagent

Quality Level

Assay

≥98%

form

crystals or chunks

impurities

≤0.01% insolubles

pH

13 (25 °C, 190.1 g/L)

density

1.62 g/mL at 25 °C

anion traces

chloride (Cl-): ≤0.001%
sulfate (SO42-): ≤0.01%

cation traces

Fe: ≤0.001%
excess alkali (as NaOH): ≤2.5%
heavy metals (as Pb): ≤0.001%

SMILES string

O.O.O.O.O.O.O.O.O.O.O.O.[Na+].[Na+].[Na+].[O-]P([O-])([O-])=O

InChI

1S/3Na.H3O4P.12H2O/c;;;1-5(2,3)4;;;;;;;;;;;;/h;;;(H3,1,2,3,4);12*1H2/q3*+1;;;;;;;;;;;;;/p-3

InChI key

ASTWEMOBIXQPPV-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Application

Sodium phosphate tribasic dodecahydrate may be used in the following processes:
  • Fabrication of chitosan membranes for use as the polymer electrolyte in a direct borohydride fuel cell (DBFC).
  • Synthesis of hierarchical silver phosphate microcrystals with photocatalytic activity via template-free method.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Evaluation of multivalent phosphate cross-linked chitosan biopolymer membrane for direct borohydride fuel cells
Ma J, et al
Journal of Power Sources, 202, 18-27 (2012)
Template-free and eco-friendly synthesis of hierarchical Ag3PO4 microcrystals with sharp corners and edges for enhanced photocatalytic activity under visible light
Kumar S, et al
Materials Letters, 123, 172-175 (2014)
Cloud point phenomenon in amphiphilic drug solutions.
Kim EJ and Shah DO.
Langmuir, 18(26), 10105-10108 (2002)
Milne GWA.
Gardner's Commercially Important Chemicals: Synonyms, Trade Names, and Properties, 573-573 (2005)
Inkyu Park et al.
Biosensors & bioelectronics, 22(9-10), 2065-2070 (2006-10-24)
A microneedle sensor platform with integrated silicon nanowire tip was developed for intracellular biochemical detection. Because of the virtue of miniaturized size and high sensitivity, this sensor has a great potential for studying individual cell or localized bioenvironment by revealing

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service