Skip to Content
Merck
All Photos(3)

Key Documents

209279

Sigma-Aldrich

Titanium(II) hydride

greener alternative

−325 mesh, 98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
TiH2
CAS Number:
Molecular Weight:
49.88
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.55

Assay

98%

form

powder

autoignition temp.

437 °F

reaction suitability

reagent type: catalyst
core: titanium

reagent type: reductant

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

particle size

−325 mesh

mp

>400 °C (dec.)

density

3.91 g/mL at 25 °C (lit.)

greener alternative category

SMILES string

[H][Ti][H]

InChI

1S/Ti.2H

InChI key

COEYXIBLSAIEKV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Find details here.

Application

Titanium(II) hydride (TiH2) is a non-stoichiometric compound that can be used in the enhancement of fracture by localized plastic deformation and brittle fracture for titanium and titanium alloy. It can be used in the hydrogen evolution and hydrogen storage. It can also be used in the foaming of aluminum due to its decomposition temperature closer to aluminum′s melting point.

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Flam. Sol. 1

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

The Mechanical Properties of Compact Titanium Produced from Titanium Hydride Powders Using Self-Propagating High-Temperature Synthesis
Dekhtyar AI, et al.
Powder Metallurgy and Metal Ceramics, 53(9-10), 549-556 (2015)
High ramp rate thermogravimetric analysis of zirconium (II) hydride and titanium (II) hydride
Licavoli JJ and Sanders PG
Thermochimica Acta, 616(43), 1-8 (2015)
Mechanical properties of titanium hydride
Xu JJ, et al.
Journal of alloys and compounds, 436(1-2), 82-85 (2007)
Hydrogen storage properties of nanosized MgH2- 0.1 TiH2 prepared by ultrahigh-energy- high-pressure milling
Lu J, et al.
Journal of the American Chemical Society, 131(43), 15843-15852 (2009)
Effect of Ti intermetallic catalysts on hydrogen storage properties of magnesium hydride
Zhou C, et al.
The Journal of Physical Chemistry, 117(25), 12973-12980 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service