Skip to Content
Merck
All Photos(3)

Documents

209201

Sigma-Aldrich

Copper(II) sulfate hydrate

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CuSO4 · xH2O
CAS Number:
Molecular Weight:
159.61 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.55

vapor pressure

7.3 mmHg ( 25 °C)

Assay

98%

form

crystalline powder

composition

Degree of hydration, 4-6

SMILES string

O.[Cu++].[O-]S([O-])(=O)=O

InChI

1S/Cu.H2O4S.H2O/c;1-5(2,3)4;/h;(H2,1,2,3,4);1H2/q+2;;/p-2

InChI key

CYKLGTUKGYURDP-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nicholas A T Irwin et al.
Current biology : CB, 31(1), 66-76 (2020-10-31)
DNA replication is a ubiquitous and conserved cellular process. However, regulation of DNA replication is only understood in a small fraction of organisms that poorly represent the diversity of genetic systems in nature. Here we used computational and experimental approaches
Min Luo et al.
Nature cell biology, 22(12), 1447-1459 (2020-11-18)
The Hippo pathway plays critical roles in cell growth, differentiation, organ development and tissue homeostasis, whereas its dysregulation can lead to tumorigenesis. YAP and TAZ are transcription co-activators and represent the main downstream effectors of the Hippo pathway. Here, we
Wei Zhao et al.
Dalton transactions (Cambridge, England : 2003), (8)(8), 1509-1517 (2005-04-13)
Three novel metal-organic frameworks (MOFs), [Cu(1)SO4].H2O (4), [Cu2(2)2(SO4)2].4H2O (5) and [Cu(3)(H2O)]SO4.5.5H2O (6), were obtained by hydrothermal reactions of CuSO4.5H2O with the corresponding ligands, which have different flexibility. The structures of the synthesized complexes were determined by single-crystal X-ray diffraction analyses.
Damijana Urankar et al.
Journal of combinatorial chemistry, 10(6), 981-985 (2008-10-17)
Azoamides, previously established as bioactive intracellular GSH-depleting agents, were decorated with a terminal alkyne moiety to 4 and then were transformed, by copper(I)-catalyzed azide-alkyne cycloaddition (CuAAC), into different ligand-arm functionalized azoamides 6. Azides 5 having ligand-arms amenable for binding to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service