Skip to Content
Merck
All Photos(1)

Key Documents

201030

Sigma-Aldrich

Osmium tetroxide

ReagentPlus®, 99.8%

Synonym(s):

Osmium(VIII)-oxide, ‘Osmic acid’

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
OsO4
CAS Number:
Molecular Weight:
254.23
EC Number:
MDL number:
UNSPSC Code:
12352303
PubChem Substance ID:
NACRES:
NA.55
grade:
reagent
form:
crystalline
powder

grade

reagent

vapor density

8.8 (vs air)

vapor pressure

7 mmHg ( 20 °C)

product line

ReagentPlus®

Assay

99.8%

form

crystalline
powder

reaction suitability

reagent type: oxidant

bp

130 °C (lit.)

mp

39.5-41 °C (lit.)

storage temp.

2-8°C

SMILES string

O=[Os](=O)(=O)=O

InChI

1S/4O.Os

InChI key

VUVGYHUDAICLFK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Osmium tetroxide (OsO4) is widely employed as a catalyst in various organic reactions. Stereochemical aspects of the OsO4 assisted oxidation of allylic alcohols and their derivatives have been reported. Its reaction with olefins has been investigated. It participates as a catalyst for oxidizing fluoroquinolones, via reaction with potassium hexacyanoferrate(III) in alkaline medium. Kinetics of this oxidation reaction was studied.

Application

Osmium tetroxide may be employed as a catalyst for the hydroxylation of sterically hindered olefins.
Osmium tetroxide may be used in the following processes:
  • To promote the lactonization of alkenols to lactones.
  • To mediate the selective oxidative cleavage of olefins by oxone to form the corresponding ketones or carboxylic acids.
  • Dihydroxylation of mono-trans-di, and trisubstituted olefins to form 1,2-diols with high enantioselectivity in the presence of chiral amines.
Widely used oxidizer for visualizing cells in microscopy. Promotes oxidative cleavage of graphene layers in carbon nanotubes creating the possibility of opening ′windows′ in fullerene cages.

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Osmium tetroxide-promoted catalytic oxidative cleavage of olefins: an organometallic ozonolysis.
Travis BR, et al.
Journal of the American Chemical Society, 124(15), 3824-3825 (2002)
Hwang, K.C.
Journal of the Chemical Society. Chemical Communications, 173-173 (1995)
Highly enantioselective dihydroxylation of olefins by osmium tetroxide with chiral diamines.
Nakajima M, et al.
Tetrahedron, 49(47), 10793-10806 (1993)
Osmium tetroxide catalyzed hydroxylation of hindered olefins.
Ray R and Matteson DS.
Tetrahedron Letters, 21(5), 449-450 (1980)
Direct Lactonization of Alkenols via Osmium Tetroxide-Mediated Oxidative Cleavage.
Schomaker JM, et al.
Organic Letters, 5(17), 3089-3092 (2003)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service