Skip to Content
Merck
All Photos(2)

Key Documents

179965

Sigma-Aldrich

Toluene

Laboratory Reagent, ≥99.3%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH3
CAS Number:
Molecular Weight:
92.14
Beilstein:
635760
EC Number:
MDL number:
UNSPSC Code:
12191501
eCl@ss:
39011102
PubChem Substance ID:
grade:
Laboratory Reagent
application(s):
microbiology
bp:
110-111 °C (lit.)
vapor pressure:
22 mmHg ( 20 °C)
26 mmHg ( 25 °C)
Pricing and availability is not currently available.

grade

Laboratory Reagent

Quality Level

vapor density

3.2 (vs air)

vapor pressure

22 mmHg ( 20 °C)
26 mmHg ( 25 °C)

Assay

≥99.3%

form

liquid

autoignition temp.

997 °F

expl. lim.

7 %

dilution

(for analytical testing)

impurities

≤0.05% water

evapn. residue

≤0.005%

refractive index

n/D 1.496 (lit.)

bp

110-111 °C (lit.)

mp

-93 °C (lit.)

density

0.865 g/mL at 25 °C (lit.)

application(s)

microbiology

SMILES string

Cc1ccccc1

InChI

1S/C7H8/c1-7-5-3-2-4-6-7/h2-6H,1H3

InChI key

YXFVVABEGXRONW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Toluene, a flammable liquid with a pungent odor, is widely employed as organic solvent. It is widely used as a precursor for synthesizing benzene and as a solvent in the paint industry.[1] It has been reported to be a biotoxic solvent (toxic to many microorganisms at 0.1%v/v concentrations).[2] Its anaerobic biodegradation to CO2, by the denitrifying bacterium Thauera arornatica has been reported.[3] It forms syndiotactic polystyrene-toluene molecular compound. Crystal structure of this molecular compound has been investigated by X-ray diffraction studies.[4]

Application

Toluene may be used as a solvent in the following studies:
  • Preparation of 4-arm polylactide-based (PLA) star oligomer.[5]
  • Transformation of C6 sugar monomers/oligomers (glucose, maltose, and β-cyclodextrins) to levulinic acid in the presence of an acid catalyst.[6]
Toluene undergoes alkylation in the presence of modified ZSM (Zeolite Socony Mobil)-5-class zeolite catalysts to form p-xylene with high selectivity.[7] When doped on graphene, it acts as an electron donor leading to alteration in graphene electrical properties.[8]

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 3 - Asp. Tox. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 2 Inhalation - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

39.9 °F - closed cup

Flash Point(C)

4.4 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 3

1 of 3

Direct laser writing of 3D scaffolds for neural tissue engineering applications.
Melissinaki V, et al.
Biofabrication, 3(4), 045005-045005 (2011)
Acid-catalyzed conversion of C6 sugar monomer/oligomers to levulinic acid in water, tetrahydrofuran and toluene: Importance of the solvent polarity.
Hu X, et al.
Fuel: The Science and Technology of Fuel and Energy, 141, 56-63 (2015)
Selective alkylation of toluene with methanol to produce para-xylene.
Kaeding WW, et al.
J. Catal., 67(1), 159-174 (1981)
Toluene.
von Burg R.
Journal of Applied Toxicology, 13(6), 441-446 (1993)
Electrochemical doping of graphene with toluene.
Kaverzin AA, et al.
Carbon, 49(12), 3829-3834 (2011)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service