Skip to Content
Merck
All Photos(1)

Key Documents

04231

Sigma-Aldrich

Calcium phosphate dibasic dihydrate

puriss., meets analytical specification of Ph. Eur., BP, USP, 98-102.5%

Synonym(s):

Calcium hydrogen phosphate dihydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CaHPO4 · 2H2O
CAS Number:
Molecular Weight:
172.09
EC Number:
MDL number:
UNSPSC Code:
51101500
PubChem Substance ID:
NACRES:
NA.21
Assay:
98-102.5%
form:
powder

grade

puriss.

Quality Level

Assay

98-102.5%

form

powder

quality

meets analytical specification of Ph. Eur., BP, USP

impurities

organic volatile impurities, complies
residual solvents, complies
≤0.003% heavy metals (as Pb)
≤0.1% insoluble in HCl

loss

24.5-26.5% loss on ignition, 800 °C

anion traces

carbonate (CO32-): in accordance
chloride (Cl-): ≤200 mg/kg
fluoride (F-): ≤50 mg/kg
sulfate (SO42-): ≤3000 mg/kg

cation traces

As: ≤1 mg/kg
Ba:, in accordance (Ph Eur)
Ba:, in accordance (USP)
Fe: ≤300 mg/kg

SMILES string

O.O.[Ca++].OP([O-])([O-])=O
O.O.[Ca++].OP([O-])([O-])=O

InChI

1S/Ca.H3O4P.2H2O/c;1-5(2,3)4;;/h;(H3,1,2,3,4);2*1H2/q+2;;;/p-2

InChI key

XAAHAAMILDNBPS-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application


  • Simultaneous Regulation of the Mechanical/Osteogenic Capacity of Brushite Calcium Phosphate Cement by Incorporating with Poly(ethylene glycol) Dicarboxylic Acid: This research investigates enhancing brushite calcium phosphate cements by adding poly(ethylene glycol) dicarboxylic acid to improve mechanical properties and osteogenic capabilities, applicable in bone regeneration and implant materials. (Gao et al., 2024).

  • Biosurfactant-Assisted Cu Doping of Brushite Coatings: Explores the structural and biofunctional enhancements of brushite coatings through copper doping facilitated by biosurfactants, which could revolutionize surface treatments for biocompatible implants and devices. (Siva Prasad et al., 2024).

  • Self-Healing Micro Arc Oxidation and Dicalcium Phosphate Dihydrate Double-Passivated Coating on Magnesium Membrane for Enhanced Bone Integration Repair: This study highlights the application of calcium phosphate dibasic dihydrate in developing protective coatings on magnesium-based implants, significantly advancing orthopedic and dental implant technologies. (Wu et al., 2024).

  • Physiologically Engineered Porous Titanium/Brushite Scaffolds for Critical-Size Bone Defects: Details the design and fabrication of novel scaffolds for treating large bone defects, utilizing a combination of titanium and brushite, highlighting its potential in reconstructive surgery and regenerative medicine. (Abdulaziz et al., 2023).

  • Mechanochemical Synthesis of Cerium Chlorapatite from a Mixture of Cerium Chloride Heptahydrate, Dicalcium Phosphate Dihydrate, and Calcium Hydroxide for Biomedical Application: Focuses on the innovative production of cerium chlorapatite using dicalcium phosphate dihydrate, showcasing its potential in biomedical engineering, especially in drug delivery and tissue engineering. (Otsuka et al., 2024).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Moreton, R. C.
Chemical & Pharmaceutical Bulletin, 868-870 (1996)
J M Lausier et al.
Journal of pharmaceutical sciences, 66(11), 1636-1637 (1977-11-01)
The aging of direct compression tablets made using dibasic calcium phosphate dihydrate as the tablet matrix was investigated over 16 weeks. The formula included 6% amaranth as a dye tracer. Two sets of stress storage conditions were used: 25 degrees
Guiling Zhang et al.
Journal of inorganic biochemistry, 113, 1-8 (2012-06-13)
Bisphosphonates (BPs) are a class of synthetic pyrophosphate analogs that can prevent the loss of bone mass, given orally to treat postmenopuasal osteoporosis. It is not clear yet if the benefits of BPs include the possibility of affecting bone apatitic
F Michaut et al.
Journal of pharmaceutical sciences, 99(6), 2874-2885 (2009-12-30)
The Stylcam 100R, a rotary press simulator, was designed to simulate speed profiles of rotary tablet presses. Such a simulator was qualified by numerous laboratories and, actually, its ability to be used for studying the behaviour of powders under pressure
Sarsvatkumar Patel et al.
International journal of pharmaceutics, 389(1-2), 66-73 (2010-01-20)
This work proposed to study the influence of varying compaction pressure on the plastic energy, elasticity (Young's modulus), particle yield strength, strain hardening, and applied pressures on derived Heckel parameter using material with different densification and deformation mechanisms: ibuprofen (IBN)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service