Skip to Content
Merck
All Photos(1)

Key Documents

Y0001563

Mesalazine impurity A

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

4-Aminophenol, 4-Hydroxyaniline

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NC6H4OH
CAS Number:
Molecular Weight:
109.13
Beilstein:
385836
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

mesalazine, mesalamine

manufacturer/tradename

EDQM

mp

185-189 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

Nc1ccc(O)cc1

InChI

1S/C6H7NO/c7-5-1-3-6(8)4-2-5/h1-4,8H,7H2

InChI key

PLIKAWJENQZMHA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Mesalazine impurity A EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Product No.
Description
Pricing

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Muta. 2 - Skin Sens. 1 - STOT RE 2

Target Organs

Kidney

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Soumabha Bag et al.
Analytica chimica acta, 860, 37-42 (2015-02-16)
Ionization of aliphatic and aromatic aldehydes is improved by performing simultaneous chemical derivatization using 4-aminophenol to produce charged iminium ions during paper spray ionization. Accelerated reactions occur in the microdroplets generated during the paper spray ionization event for the tested
Chongming Liu et al.
Journal of mass spectrometry : JMS, 49(8), 692-699 (2014-07-22)
On contrary to the widely accepted conviction that the m/z 93 ion derived from phenol does not react with CO2, we demonstrate that it makes an adduct with CO2 to a small but demonstrable extent. For example, the product-ion mass
Denial Mahata et al.
International journal of biological macromolecules, 69, 5-11 (2014-05-20)
Cardanol is a non-isoprenoic phenolic lipid-mixture of distilled cashew nut shell liquid obtained from Anacardium occidentale. Herein, cardanol is purified from cashew nut shell liquid (CNSL) and synthesized to new compounds with different azo amphiphiles. These synthesized compounds are allowed
Ahmad Aqel et al.
Journal of chromatographic science, 52(3), 201-210 (2013-02-21)
This paper describes the comprehensive fabrication of monolithic materials for use as stationary phases in capillary liquid chromatography. Several columns were synthesized in the confines of 320 µm i.d. fused-silica capillaries by single-step in situ copolymerization of benzyl methacrylate and
Paula Paíga et al.
Journal of pharmaceutical and biomedical analysis, 106, 61-70 (2014-07-09)
An analytical methodology for the simultaneous determination of seven pharmaceuticals and two metabolites belonging to the non-steroidal anti-inflammatory drugs (NSAIDs) and analgesics therapeutic groups was developed based on off-line solid-phase extraction and ultra-high performance liquid chromatography coupled to tandem mass

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service