Skip to Content
Merck
All Photos(1)

Key Documents

Y0001448

Bicalutamide for system suitability

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Bicalutamide (CDX), Casodex, Cosudex, N-[4-Cyano-3-(trifluoromethyl)phenyl]-3-[(4-fluorophenyl)sulfonyl]-2-hydroxy-2-methylpropanamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H14F4N2O4S
CAS Number:
Molecular Weight:
430.37
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

bicalutamide

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

CC(O)(CS(=O)(=O)c1ccc(F)cc1)C(=O)Nc2ccc(C#N)c(c2)C(F)(F)F

InChI

1S/C18H14F4N2O4S/c1-17(26,10-29(27,28)14-6-3-12(19)4-7-14)16(25)24-13-5-2-11(9-23)15(8-13)18(20,21)22/h2-8,26H,10H2,1H3,(H,24,25)

InChI key

LKJPYSCBVHEWIU-UHFFFAOYSA-N

Gene Information

human ... AR(367)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Bicalutamide for system suitability EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Bicalutamide (CDX) is a non-steriodal Androgen Receptor (AR) Antagonist and a pure antiandrogen.
Bicalutamide (CDX) is a non-steriodal Androgen Receptor (AR) antagonist and a pure antiandrogen. It acts via balancing histone acetylation/deacetylation and recruitment of coregulators. Bicalutamide (CDX) abolishes androgen-mediated expression. For example, MMP13 upregulation in prostate cancer, PLZF (promyelocytic leukemia zinc finger protein), and GADD45γ (growth arrest and DNA damage inducible, gamma). Bicalutamide (CDX) is inhibited by non-genomic, transcription-independent stimulation of PI3K/AKT phosphorylation by androgens.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Health hazardEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 1 - Carc. 2 - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Peter Iversen
The Journal of urology, 170(6 Pt 2), S48-S52 (2003-11-12)
The current evidence is considered to support 150 mg of the nonsteroidal antiandrogen bicalutamide for early stage prostate cancer. Data from phase III trials of 150 mg bicalutamide monotherapy for locally advanced disease are discussed. In addition, the first overall
Ian D Cockshott
Clinical pharmacokinetics, 43(13), 855-878 (2004-10-29)
Bicalutamide is a nonsteroidal pure antiandrogen given at a dosage of 150 mg once daily as monotherapy for the treatment of early (localised or locally advanced) nonmetastatic prostate cancer. It is used at a dosage of 50 mg once daily
Keri Wellington et al.
Drugs, 66(6), 837-850 (2006-05-19)
Bicalutamide (Casodex) is a competitive androgen receptor antagonist that inactivates androgen-regulated prostate cell growth and function, leading to cell apoptosis and inhibition of prostate cancer growth. It is administered orally as a once-daily dose. In the EU and a number
Bicalutamide ('Casodex') development: from theory to therapy.
G J Kolvenbag et al.
The cancer journal from Scientific American, 3(4), 192-203 (1997-07-01)
Manfred P Wirth et al.
Frontiers of radiation therapy and oncology, 41, 39-48 (2008-06-12)
Several randomized trials have demonstrated that adjuvant medical or surgical castration may improve overall survival in patients with locally advanced prostate cancer undergoing external beam radiotherapy. After radical prostatectomy, patients with positive lymph nodes seem to benefit from adjuvant hormonal

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service