Skip to Content
Merck
All Photos(1)

Key Documents

Y0001165

Tetrandrine

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

(1β)-6,6′,7,12-Tetramethoxy-2,2′-dimethylberbaman

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C38H42N2O6
CAS Number:
Molecular Weight:
622.75
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

tetrandrine

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

COc1ccc2C[C@@H]3N(C)CCc4cc(OC)c(OC)c(Oc5cc6[C@H](Cc7ccc(Oc1c2)cc7)N(C)CCc6cc5OC)c34

InChI

1S/C38H42N2O6/c1-39-15-13-25-20-32(42-4)34-22-28(25)29(39)17-23-7-10-27(11-8-23)45-33-19-24(9-12-31(33)41-3)18-30-36-26(14-16-40(30)2)21-35(43-5)37(44-6)38(36)46-34/h7-12,19-22,29-30H,13-18H2,1-6H3/t29-,30-/m0/s1

InChI key

WVTKBKWTSCPRNU-KYJUHHDHSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Tetrandrine EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Huimin Zou et al.
International immunopharmacology, 69, 307-312 (2019-02-16)
Tetrandrine (TET) is an anti-inflammatory compound isolated from Chinese herb Stephania tetrandra S. Moore. It was reported recently that the differentiation of Th17 cells was inhibited, while the generation of induced Treg cells (iTregs) was promoted, by TET treatment. We
Ke Wu et al.
International journal of oncology, 46(3), 1205-1213 (2014-12-20)
Colon cancer is one of the most common malignancies, causes considerable morbidity and mortality. The current treatment for colon cancer is more modest than had been hoped. There is an urgent clinical need to explore new agents or adjuvants for
Chiu-Yin Kwan et al.
Acta pharmacologica Sinica, 23(12), 1057-1068 (2002-12-06)
Tetrandrine (TET), a bis-benzylisoquinoline alkaloid purified and identified an active ingredient in a Chinese medicinal herb, radix stephanae tetrandrae, has been used traditionally for the treatment of congestive circulatory disorder and inflammatory diseases. TET, together with a few of its
Yu-Jen Chen
Acta pharmacologica Sinica, 23(12), 1102-1106 (2002-12-06)
Tetrandrine, a bisbenylisoquinoline alkaloid isolated from the dried root of Stephenia tetrandra S Moore, exhibits very broad pharmacological actions, including anti-tumor activity. The beneficial effects of tetrandrine on tumor cell cytotoxicity and radiosensitization, multidrug resistance, normal tissue radioprotection, and angiogenesis
Pharmacology of tetrandrine and its therapeutic use in digestive diseases.
D G Li et al.
World journal of gastroenterology, 7(5), 627-629 (2002-01-31)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service