Skip to Content
Merck
All Photos(1)

Documents

Y0001146

Cimetidine for system suitability

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Cimetidine, SKF-92334

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H16N6S
CAS Number:
Molecular Weight:
252.34
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

cimetidine

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

−20°C

SMILES string

CN\C(NC#N)=N\CCSCc1nc[nH]c1C

InChI

1S/C10H16N6S/c1-8-9(16-7-15-8)5-17-4-3-13-10(12-2)14-6-11/h7H,3-5H2,1-2H3,(H,15,16)(H2,12,13,14)

InChI key

AQIXAKUUQRKLND-UHFFFAOYSA-N

Gene Information

human ... HRH2(3274)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Cimetidine for system suitability EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J P Harcourt et al.
International journal of pediatric otorhinolaryngology, 51(2), 109-113 (2000-01-05)
Effective adjuvant treatment for recurrent respiratory papillomatosis (RRP) is at present limited to alpha-interferon, which may have significant side effects including rebound growth of papillomata following its withdrawal, is given by injection and is expensive. High dose cimetidine is known
A Kumar
DICP : the annals of pharmacotherapy, 24(3), 289-295 (1990-03-01)
Suppressor T lymphocytes possess histamine2 (H2) receptors and contribute significantly to the function of the immune system. Experimentally, cimetidine, an H2-receptor antagonist, has been shown to enhance a variety of immunologic functions both in vivo and in vitro because of
P San Miguel et al.
Virchows Archiv : an international journal of pathology, 430(3), 261-263 (1997-03-01)
A case of carcinoma in the breast of a male patient with a chronic gastric ulcer is described. The patient had received cimetidine for 17 years. Histological examination of an excisional biopsy showed lobular in situ and infiltrating carcinoma. In
Cimetidine in idiopathic urticaria.
M Choy et al.
DICP : the annals of pharmacotherapy, 25(6), 609-612 (1991-06-01)
A F Shinn
Drug safety, 7(4), 245-267 (1992-07-01)
The excellent efficacy and tolerability profiles of H2-antagonists have established these agents as the leading class of antiulcer drugs. Attention has been focused on drug interactions with H2-antagonists as a means of product differentiation and because many patients are receiving

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service