Skip to Content
Merck
All Photos(1)

Key Documents

Y0001104

Picosulfate for system suitability

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Sodium picosulfate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H13NNa2O8S2
CAS Number:
Molecular Weight:
481.41
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

sodium picosulfate

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

[Na+].[Na+].[S](=O)(=O)([O-])Oc1ccc(cc1)C(c3ncccc3)c2ccc(cc2)O[S](=O)(=O)[O-]

InChI

1S/C18H15NO8S2.2Na/c20-28(21,22)26-15-8-4-13(5-9-15)18(17-3-1-2-12-19-17)14-6-10-16(11-7-14)27-29(23,24)25;;/h1-12,18H,(H,20,21,22)(H,23,24,25);;/q;2*+1/p-2

InChI key

GOZDTZWAMGHLDY-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Picosulfate for system suitability EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Annie O O Chan
Hong Kong medical journal = Xianggang yi xue za zhi, 18(5), 388-394 (2012-09-29)
OBJECTIVES. Although Picolax (sodium picosulphate and magnesium citrate) has been widely documented for use in bowel preparation, there is limited literature on its efficacy in the treatment of constipation. Refractory constipation is a more difficult situation with limited treatment options
Drug eruption due to sodium picosulfate.
Asuka Ishikawa et al.
European journal of dermatology : EJD, 22(3), 410-411 (2012-03-13)
Jennifer A Flemming et al.
Gastrointestinal endoscopy, 75(3), 537-544 (2011-12-24)
Picosulfate, magnesium oxide, and citric acid solution is a small-volume agent for colon cleansing before colonoscopy that is extremely well tolerated by patients, safe, and efficacious. Studies of other cleansing agents have suggested that split-dose regimens may further enhance efficacy.
G Manes et al.
Colorectal disease : the official journal of the Association of Coloproctology of Great Britain and Ireland, 15(9), 1145-1153 (2013-04-16)
The study compared the efficacy, safety and tolerability of a low-volume picosulphate/magnesium citrate preparation with that of polyethylene glycol plus ascorbic acid (PEG + ASC) in a randomized clinical trial (RCT). A multicentre randomized, single-blinded study was designed. Adult outpatients
A Slater et al.
The British journal of radiology, 85(1016), e410-e415 (2011-12-15)
The aim of this study was to determine if the introduction of faecal tagging to CT colonography (CTC) made the examination easier to tolerate or reduced the number of false-positives. Our department changed bowel preparation for CT colonography from Picolax

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service