Skip to Content
Merck
All Photos(1)

Key Documents

Y0001090

Fluvastatin sodium

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

(3R,5S,6E)-rel-7-[3-(4-Fluorophenyl)-1-(1-methylethyl)-1H-indol-2-yl]-3,5-dihydroxy-6-heptenoic acid monosodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H25FNNaO4
CAS Number:
Molecular Weight:
433.45
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

fluvastatin

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

[Na+].CC(C)n1c(\C=C\[C@H](O)C[C@H](O)CC([O-])=O)c(-c2ccc(F)cc2)c3ccccc13

InChI

1S/C24H26FNO4.Na/c1-15(2)26-21-6-4-3-5-20(21)24(16-7-9-17(25)10-8-16)22(26)12-11-18(27)13-19(28)14-23(29)30;/h3-12,15,18-19,27-28H,13-14H2,1-2H3,(H,29,30);/q;+1/p-1/b12-11+;/t18-,19-;/m0./s1

InChI key

ZGGHKIMDNBDHJB-RPQBTBOMSA-M

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Fluvastatin sodium EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Keith B Hoffman et al.
PloS one, 7(8), e42866-e42866 (2012-09-01)
Cholesterol management drugs known as statins are widely used and often well tolerated; however, a variety of muscle-related side effects can arise. These adverse events (AEs) can have serious impact, and form a significant barrier to therapy adherence. Surveillance of
Janine Kah et al.
Oncology reports, 28(3), 1077-1083 (2012-06-20)
HMG-CoA-reductase inhibitors (statins) are widely used drugs to interfere with cholesterol biosynthesis. Besides this usage, evidence is increasing that statins might also be useful in therapy of viral infections or cancer. We investigated the effects of fluva-, simva-, atorva-, rosuva-
Kenichiro Kinouchi et al.
Journal of atherosclerosis and thrombosis, 20(3), 245-256 (2012-12-01)
Statins not only reduce low-density lipoprotein (LDL) cholesterol, but also prevent the progression of kidney dysfunction. Ezetimibe, a cholesterol-absorption inhibitor, also lowers LDL cholesterol levels when added to statins; however, the effect of add-on ezetimibe on kidney function has had
Matthew Psaila et al.
Therapeutic advances in respiratory disease, 6(4), 243-246 (2012-07-05)
Statins are widely used for primary and secondary prevention of cardiovascular disease. For this reason, knowledge of the side effects and interactions pertaining to this class of pharmaceuticals is of utmost importance to all physicians. In this text a case
Allen B Williams et al.
Blood, 120(15), 3069-3079 (2012-08-29)
FLT3 is frequently mutated in acute myeloid leukemia (AML), but resistance has limited the benefit of tyrosine kinase inhibitors (TKI). We demonstrate that statins can impair FLT3 glycosylation, thus leading to loss of surface expression and induction of cell death

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service