Skip to Content
Merck
All Photos(1)

Key Documents

Y0000532

Sultamicillin for peak identification

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Sultamicillin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H30N4O9S2
CAS Number:
Molecular Weight:
594.66
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

sultamicillin

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

[S]1(=O)(=O)C2N([C@H](C1(C)C)C(=O)OCOC(=O)[C@@H]3N4C(SC3(C)C)C(C4=O)NC(=O)[C@@H](N)c5ccccc5)C(=O)C2

InChI

1S/C25H30N4O9S2/c1-24(2)17(29-20(32)16(21(29)39-24)27-19(31)15(26)12-8-6-5-7-9-12)22(33)37-11-38-23(34)18-25(3,4)40(35,36)14-10-13(30)28(14)18/h5-9,14-18,21H,10-11,26H2,1-4H3,(H,27,31)/t14?,15-,16?,17-,18-,21?/m0/s1

InChI key

OPYGFNJSCUDTBT-CUUJHUNTSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Sultamicillin for peak identification EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tahir Bezgin et al.
Cardiovascular journal of Africa, 24(2), e10-e12 (2012-11-16)
The concurrence of acute coronary syndrome with allergy or hypersensitivity as well as with anaphylactic or anaphylactoid reactions is increasingly encountered in daily clinical practice. There are several reports associating mast cell activation with acute cardiovascular events in adults. This
Mahir Seyrek et al.
Journal of endourology, 26(11), 1431-1436 (2012-05-23)
To compare sulbactam-ampicillin and cefuroxime antibiotics for prophylaxis of percutaneous nephrolithotomy (PCNL) and to find out the optimal regimen for antibiotic maintenance to prevent systemic inflammatory response syndrome (SIRS). Between February 2010 and March 2011, a total of 198 patients
Mesut Yilmaz et al.
European journal of gastroenterology & hepatology, 24(6), 688-694 (2012-03-22)
Antibiotic-associated hemorrhagic colitis is a distinct form of antibiotic-associated bloody diarrhea (AABD) in which Clostridium difficile is absent. Although the cause is not exactly known, reports have suggested the role of Klebsiella oxytoca and/or C. difficile. Between 2001 and 2006
Hiroki Tsukada et al.
Journal of infection and chemotherapy : official journal of the Japan Society of Chemotherapy, 18(5), 715-721 (2012-03-31)
Klebsiella pneumoniae is an important causative bacterium of aspiration pneumonia in many elderly patients. We retrospectively investigated the clinical effects of the early treatment of aspiration pneumonia and background factors in 24 patients from whom Klebsiella pneumoniae was isolated. Sulbactam/ampicillin
Norhafiza Mat Lazim et al.
International journal of pediatric otorhinolaryngology, 77(4), 457-461 (2013-01-01)
Tonsillectomy is a common operative procedure performed for tonsillar hypertrophy complicates with recurrent tonsillitis. Among the post tonsillectomy morbidities, post operative wound healing is of utmost importance to be effectively managed as it will interfere with patient recuperation from surgery.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service