Select a Size
| Size/SKU | Availability | Price |
|---|
About This Item
grade
certified reference material
shelf life
limited shelf life, expiry date on the label
manufacturer/tradename
Paragon Scientific Ltd
ASTM D 2162
12423 mPa.s(25.00 °C)
application(s)
cosmetics
flavors and fragrances
food and beverages
oil and petrochemicals
personal care
format
mixture
General description
1 of 1
This Item | |||
|---|---|---|---|
| format mixture | format mixture | format mixture | format mixture |
| grade certified reference material | grade certified reference material | grade certified reference material | grade certified reference material |
| application(s) cosmetics | application(s) cosmetics | application(s) cosmetics | application(s) cleaning products |
| manufacturer/tradename Paragon Scientific Ltd | manufacturer/tradename Paragon Scientific Ltd | manufacturer/tradename Paragon Scientific Ltd | manufacturer/tradename Paragon Scientific Ltd |
| shelf life limited shelf life, expiry date on the label | shelf life limited shelf life, expiry date on the label | shelf life limited shelf life, expiry date on the label | shelf life limited shelf life, expiry date on the label |
| ASTM D 2162 12423 mPa.s(25.00 °C) | ASTM D 2162 4.988 mPa.s(25.00 °C) | ASTM D 2162 251.5 mPa.s(25.00 °C) | ASTM D 2162 30036 mPa.s(25.00 °C) |
Still not finding the right product?
Explore all of our products under Silicone Rotational Viscosity Standard
Storage Class
10 - Combustible liquids
wgk
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
