Select a Size
| Size/SKU | Availability | Price |
|---|
About This Item
grade
certified reference material
CofA
current certificate can be downloaded
packaging
pkg of 1.5 mL
manufacturer/tradename
RTC UST-148
concentration
50,000 μg/mL in methylene chloride
technique(s)
HPLC: suitable, gas chromatography (GC): suitable
application(s)
environmental
petroleum
format
matrix material
storage temp.
2-30°C
General description
Application
Other Notes
1 of 1
This Item | |||
|---|---|---|---|
| technique(s) HPLC: suitable, gas chromatography (GC): suitable | technique(s) HPLC: suitable, gas chromatography (GC): suitable | technique(s) HPLC: suitable, gas chromatography (GC): suitable | technique(s) HPLC: suitable, gas chromatography (GC): suitable |
| format matrix material | format matrix material | format matrix material | format matrix material |
| Quality Level 100, 300 | Quality Level 100, 300 | Quality Level 100, 300 | Quality Level 100, 300 |
| grade certified reference material | grade certified reference material | grade certified reference material | grade certified reference material |
| storage temp. 2-30°C | storage temp. 2-30°C | storage temp. 2-30°C | storage temp. 2-30°C |
| packaging pkg of 1.5 mL | packaging pkg of 1.5 mL | packaging - | packaging pkg of 1.5 mL |
signalword
Warning
hcodes
Hazard Classifications
Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
target_organs
Central nervous system
Storage Class
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
wgk
WGK 2
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.





