Skip to Content
Merck
All Photos(1)

Key Documents

T0040800

Tenoxicam

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

4-Hydroxy-2-methyl-N-2-pyridinyl-2H-thieno(2,3-e)-1,2-thiazine-3-carboxamide 1,1-dioxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H11N3O4S2
CAS Number:
Molecular Weight:
337.37
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

tenoxicam

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

O=C(NC1=NC=CC=C1)C2=C(O)C(SC=C3)=C3S(N2C)(=O)=O

InChI

1S/C13H11N3O4S2/c1-16-10(13(18)15-9-4-2-3-6-14-9)11(17)12-8(5-7-21-12)22(16,19)20/h2-7,17H,1H3,(H,14,15,18)

InChI key

LZNWYQJJBLGYLT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Tenoxicam EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Non-steroidal antiinflammatory drug (NSAID) with comparatively low risk of renal or hepatic toxicity.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michael H Bennett et al.
The Cochrane database of systematic reviews, 5(5), CD005277-CD005277 (2012-05-18)
Decompression illness (DCI) is due to bubble formation in the blood or tissues following the breathing of compressed gas. Clinically, DCI may range from a trivial illness to loss of consciousness, death or paralysis. Recompression is the universally accepted standard
Photoclinic. Fixed drug eruption.
Vitorino Modesto Santos et al.
Archives of Iranian medicine, 14(1), 71-72 (2011-01-05)
Marco Franco-Pérez et al.
The journal of physical chemistry. B, 115(46), 13593-13598 (2011-10-11)
The determination of the micro-equilibrium prototropic constants is often a tough task when the tautomeric ratio favors one of the species or when the chemical exchange is not slow enough to allow the quantitative detection of the tautomeric species. There
Owen A Moore et al.
The Cochrane database of systematic reviews, (3)(3), CD007591-CD007591 (2009-07-10)
Tenoxicam is a non-steroidal anti-inflammatory drug (NSAID) licensed for use in rheumatic disease and other musculoskeletal disorders in the UK, and is widely available in other countries worldwide. This review sought to evaluate the efficacy and safety of oral tenoxicam
Sutapa Mondal Roy et al.
Langmuir : the ACS journal of surfaces and colloids, 27(24), 15054-15064 (2011-10-18)
Membrane fusion is an essential process guiding many important biological events, which most commonly requires the aid of proteins and peptides as fusogenic agents. Small drug induced fusion at low drug concentration is a rare event. Only three drugs, namely

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service