Skip to Content
Merck
All Photos(1)

Key Documents

SMB01382

Sigma-Aldrich

2-Methoxyestrone

≥95% (HPLC), from synthetic, solid

Synonym(s):

2,3-Dihydroxy-1,3,5(10)-estratrien-17-one 2-methyl ether, 2-Hydroxyestrone 2-methyl ether, 3-Hydroxy-2-methoxy-1,3,5(10)-estratrien-17-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H24O3
CAS Number:
Molecular Weight:
300.39
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.79

biological source

synthetic

Quality Level

Assay

≥95% (HPLC)

form

solid

mp

187-190 °C

storage temp.

2-8°C

SMILES string

C[C@@]12[C@](CCC2=O)([H])[C@]3([H])CCC4=CC(O)=C(OC)C=C4[C@@]3([H])CC1

InChI

1S/C19H24O3/c1-19-8-7-12-13(15(19)5-6-18(19)21)4-3-11-9-16(20)17(22-2)10-14(11)12/h9-10,12-13,15,20H,3-8H2,1-2H3/t12-,13+,15-,19-/m0/s1

InChI key

WHEUWNKSCXYKBU-QPWUGHHJSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-Methoxyestrone is an endogenous, naturally occurring methoxylated catechol estrogen and an estrone metabolite formed by catechol O-methyltransferase via the intermediate 2-hydroxyestrone. Unlike estrone but similarly to 2-hydroxyestrone and 2-methoxyestradiol, 2-methoxyestrone has very low affinity for the estrogen receptor and lacks significant estrogenic activity

Storage and Stability

Heat sensitive

Pictograms

Health hazardEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Urinary estrogen metabolites and gastric cancer risk among postmenopausal women.
Camargo MC, et al.
Cancer reports (Hoboken, N.J.), 5, e1574-e1574 (2022)
Individual and cyclic estrogenic profile in women: Structure and variability of the data.
Bozzolino C, et al.
Steroids, 150, 108432-108432 (2019)
Reproducibility of fifteen urinary estrogens and estrogen metabolites over a 2- to 3-year period in premenopausal women.
Eliassen AH, et al.
Cancer Epidemiology, Biomarkers & Prevention, 18, 2860-2868 (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service