Skip to Content
Merck
All Photos(1)

Key Documents

SMB01376

Sigma-Aldrich

2AA-labeled Dextran Ladder

Synonym(s):

2-Aminobenzoic acid dextran ladder standard, Glycan Ladder

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.79

Assay

>90% (HPLC)

Quality Level

form

powder or crystals

technique(s)

HPLC: suitable
mass spectrometry (MS): suitable

impurities

≤10% Unlabeled Dextran ladder

shipped in

2-8°C

storage temp.

−20°C

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service