Skip to Content
Merck
All Photos(3)

Key Documents

S9679

Sigma-Aldrich

Starch from corn

Synonym(s):

Corn starch, Starch from maize

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H10O5)n
CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.21
Pricing and availability is not currently available.

biological source

corn

Quality Level

form

solid

color

white

useful pH range

4.0 -7.0

storage temp.

room temp

Looking for similar products? Visit Product Comparison Guide

General description

Starch is a naturally occurring, abundantly available, and biodegradable polysaccharide. It is stored in the various parts of the plant such as roots, leaves, stems, tubers, fruits, and pollen grains. Corn starch is made up of 25% amylose and 75% amylopectin.[1]

Application

Starch from corn has been used for the evaluation of amylose content in bananas for standard curve preparation.[2][3] It has also been used in hydrolysis studies via in vitro fermentation using pig fecal inoculum.[4]

Biochem/physiol Actions

Starch facilitates the storage of energy in plants. Corn starch has a wide variety of applications like paper bags, stationery, tissues, manufacturing antibiotics, and drip-feed systems. It can also be used in increasing the stiffness of the fabric and improve its appearance. Corn starch is also a part of human and animal diets.[1]

Other Notes

To gain a comprehensive understanding of our extensive range of Polysaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G Giuberti et al.
Animal : an international journal of animal bioscience, 7(9), 1446-1453 (2013-06-21)
The need to improve the knowledge of fermentation processes within the digestive tract in pigs is growing, particularly for ingredients that may act as potential prebiotic sources, such as resistant starch (RS). A study (based on enzymatic digestion followed by
Effect of cryo-milling on starches: Functionality and digestibility
Dhital S, et al.
Food Hydrocolloids, 24(2-3), 152-163 (2010)
Shazia Tabasum et al.
International journal of biological macromolecules, 122, 969-996 (2018-10-21)
Maize or corn is considered as very distinctive plant. Corn having better capability of utilizing sun light, is a noble way of getting a natural polymer known as starch. Amylopectin and amylase composition in the starch firmly affects the properties
Physicochemical characteristics and resistant starch of different varieties of banana from Taiwan
Li M C, et al.
International journal of food properties, 23(1), 1168-1175 (2020)
D Dan Ramdath et al.
Foods (Basel, Switzerland), 9(2) (2020-02-15)
Proximate composition and starch nutritional properties of twenty cooked lentils were assessed to identify unique varieties that could be used in value added foods. Significant variations exist among the lentil varieties (p < 0.05) with respect to their energy, fat

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service