Skip to Content
Merck
All Photos(1)

Documents

S200

General Purpose Viscosity Standard; UKAS ISO/IEC17025 and ISO 17034 certified

viscosity 392 mPa.s (25 °C)

Synonym(s):

Paragon General Viscosity Standards

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Paragon Scientific Ltd

viscosity

392 mPa.s(25 °C)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

Looking for similar products? Visit Product Comparison Guide

General description

Please note that the value stated on this page is nominal. All certified values for the current lots of all Paragon General Viscosity Standards can be found on the Paragon Scientific webpage
This product is a certified reference material (CRM) certified to International Standards BS EN ISO / IEC 17025 and ISO 17034 under UKAS accreditation.

Application

  • Ion chromatography calibration: Utilizing a sulfate standard for IC helps in the precise calibration of ion chromatography equipment, which is essential for the accurate measurement of sulfate ions in various biological and environmental samples. This calibration is vital for studies in biochemistry where sulfate levels impact research outcomes.

Pictograms

Exclamation markHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Asp. Tox. 1 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yaaser Q Almulaiky et al.
International journal of biological macromolecules, 133, 767-774 (2019-04-21)
A novel plant peroxidase was isolated from the stem of Arabian balsam (Commiphora gileadensis) and purified using ammonium sulfate, followed by ion exchange chromatography (DEAE-Sepharose) and gel filtration (Sephcryl S-200). The newly isolated peroxidase was characterized as having a specific
Jyoti Gupta et al.
Scientific reports, 6, 39198-39198 (2016-12-17)
Bispecific antibodies (BsAbs), with the ability to recognize two different epitopes simultaneously, offer remarkable advantages in bioassays, cancer therapy, biosensors, and enzyme electrodes. Preparation and purification of BsAbs in adequate quantities remains a major hurdle in their use in various
Kavyashree Sakharayapatna Ranganatha et al.
International journal of biological macromolecules, 138, 1044-1055 (2019-07-28)
In the present study, out of three isoforms of α-mannosidase identified in the crude extract of defatted Custard apple seed powder, isoform III has been purified to homogeneity by two-step chromatography: hydrophobic interaction and gel filtration. The purified Custard apple
Naveena B Maheswarappa et al.
Proteome science, 14, 18-18 (2016-11-29)
Myoglobin (Mb) is a sarcoplasmic heme protein primarily responsible for meat color and its chemistry is species specific. 4-hydroxy-2-nonenal (HNE) is a cytotoxic lipid derived aldehyde detected in meat and was reported to covalently adduct with nucleophilic histidine residues of
Christoph Kreer et al.
Oncotarget, 8(4), 6857-6872 (2016-12-31)
N-glycosylation is generally accepted to enhance the immunogenicity of antigens because of two main reasons. First, the attachment of glycans enables recognition by endocytic receptors like the mannose receptor (MR) and hence increased uptake by dendritic cells (DCs). Second, foreign

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service