Skip to Content
Merck
All Photos(1)

Key Documents

S1340000

Stearic acid

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

1-Heptadecanecarboxylic acid, C18:0, Cetylacetic acid, NSC 25956, NSC 261168, Octadecanoic acid, Stearophanic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)16COOH
CAS Number:
Molecular Weight:
284.48
Beilstein:
608585
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

pharmaceutical primary standard

vapor pressure

1 mmHg ( 173.7 °C)

API family

stearic acid

manufacturer/tradename

EDQM

bp

361 °C (lit.)

mp

67-72 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

CCCCCCCCCCCCCCCCCC(O)=O

InChI

1S/C18H36O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18(19)20/h2-17H2,1H3,(H,19,20)

InChI key

QIQXTHQIDYTFRH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Stearic acid EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Piret Arukuusk et al.
Biochimica et biophysica acta, 1828(5), 1365-1373 (2013-01-30)
Harnessing of a branched structure is a novel approach in the design of cell-penetrating peptides and it has provided highly efficient transfection reagents for intracellular delivery of nucleic acids. The new stearylated TP10 analogs, NickFects, condense plasmid DNA, splice correcting
Qian Wan et al.
Nanoscale, 5(2), 744-752 (2012-12-12)
As a versatile gene vector, minicircle DNA (mcDNA) has a great potential for gene therapy. However, some serious challenges remain, such as to effectively deliver mcDNA into targeted cells/tissues and to non-invasively monitor the delivery of the mcDNA. Superparamagnetic iron
T A Doran et al.
American journal of obstetrics and gynecology, 133(3), 302-307 (1979-02-01)
Lecithin/sphingomyelin (L/S) ratio, creatinine, percentage of lipid-positive cells, palmitic acid, palmitic acid/stearic acid (P/S) ratio, and total cortisol were analyzed as tests for fetal maturity and fetal pulmonary maturity in 164 samples of amniotic fluid from 121 patients. Fifty samples
Wonsik Lee et al.
The Journal of biological chemistry, 288(10), 6788-6800 (2013-01-12)
Recent data indicate that the nutrients available to Mycobacterium tuberculosis (Mtb) inside its host cell are restricted in their diversity. Fatty acids and cholesterol appear more favored; however, their degradation can result in certain metabolic stresses. Their breakdown can generate
Ulla Hoppu et al.
Journal of pediatric gastroenterology and nutrition, 41(3), 335-338 (2005-09-01)
Breast milk fatty acids may have immunomodulatory properties related to the development of atopic disease. The aim of this study was to assess the impact of the breast milk fatty acid composition on the development of atopic dermatitis (AD) in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service