S0360090
(RS)-Selegiline hydrochloride
European Pharmacopoeia (EP) Reference Standard
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C13H17N · HCl
CAS Number:
Molecular Weight:
223.74
UNSPSC Code:
41116107
NACRES:
NA.24
Recommended Products
grade
pharmaceutical primary standard
API family
selegiline
manufacturer/tradename
EDQM
application(s)
pharmaceutical (small molecule)
format
neat
storage temp.
2-8°C
SMILES string
Cl.N(C(Cc1ccccc1)C)(CC#C)C
InChI
1S/C13H17N.ClH/c1-4-10-14(3)12(2)11-13-8-6-5-7-9-13;/h1,5-9,12H,10-11H2,2-3H3;1H
InChI key
IYETZZCWLLUHIJ-UHFFFAOYSA-N
General description
This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.
Application
(RS)-Selegiline hydrochloride EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.
Packaging
The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.
Other Notes
Sales restrictions may apply.
related product
Product No.
Description
Pricing
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Thomas Müller et al.
Journal of neural transmission (Vienna, Austria : 1996), 120(5), 761-765 (2012-12-01)
The objective of this study is to demonstrate that application of rasagiline instead of selegiline with concomitant determination of L-amphetamine and L-methamphetamine in plasma is safe and well tolerated and influences sleep, mood, and motor behavior in patients with Parkinson's
Qing Zhao et al.
International journal of molecular medicine, 32(4), 883-891 (2013-07-24)
The monoamine oxidase type-B (MAO-B) inhibitor, selegiline, is often recommended as a first-line treatment for Parkinson's disease (PD) and has been shwon to possess neuroprotective effects. The aim of the present study was to determine whether selegiline increases the levels
Johanna E Kowalko et al.
Current biology : CB, 23(19), 1874-1883 (2013-09-17)
Surface populations of Astyanax mexicanus, living in rivers like their common ancestors, school, while several, independently derived cave populations of the same species have lost schooling behavior. We quantify schooling behavior in individual A. mexicanus and identify quantitative trait loci (QTL)
Tamás Tábi et al.
Expert opinion on drug metabolism & toxicology, 9(5), 629-636 (2013-03-20)
Parkinson's disease affects a growing number of people. Its major treatment, levodopa therapy, however suffers from response fluctuations in the advanced phase of the disease. Adjunctive therapies have thus gained significant importance. The selective monoamine oxidase B inhibitor, selegiline
José A G Agúndez et al.
Expert opinion on drug metabolism & toxicology, 9(7), 859-874 (2013-04-10)
The development of pharmacogenetic-based clinical practice guidelines for the use of anti-Parkinson's disease drugs requires, as a pre-requisite, the identification and validation of genetic biomarkers. These biomarkers are then used as surrogate endpoints. This review analyzes potential genetic biomarkers which
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service