Skip to Content
Merck

RTM4

Mineral Oil Rotational Viscosity Standard

viscosity 2.144 mPa.s (25.00 °C)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.24
UNSPSC Code:
41116107
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

certified reference material

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Paragon Scientific Ltd

viscosity

2.144 mPa.s(25.00 °C)

application(s)

cleaning products
cosmetics
cosmetics
flavors and fragrances
food and beverages
personal care

format

mixture

General description

Mineral Oil Rotational Viscosity Standards (viscosity 2.144 mPa.s) are specifically formulated for the calibration and verification of rotational viscometers where operating temperature is controlled precisely.

Features and Benefits

  • Tested and certified at 25°C
  • Tamper-evident security packaging
  • 500mL quantity supplied in ready-to-use "test in pack" container with tamper evident cap provides absolute assurance of sample integrity
  • Full traceability to national standards
  • 1-year shelf life



Still not finding the right product?


signalword

Danger

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Skin Irrit. 2 - STOT SE 3

target_organs

Central nervous system

Storage Class

10 - Combustible liquids

wgk

WGK 1

flash_point_f

168.8 °F

flash_point_c

76 °C



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
RTM4-500ML04061837681615