Skip to Content
Merck

Skip To

RDD033

Polyvinylpyrrolidone

free-flowing, Redi-Dri™, powder, average mol wt 40,000

Synonym(s):

PVP, Polyvidone, Povidone

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Linear Formula:
(C6H9NO)n
CAS Number:
UNSPSC Code:
12352005
NACRES:
NA.25
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

product line

Redi-Dri™

form

powder

mol wt

average mol wt 40,000

SMILES string

C=CN1CCCC1=O

InChI

1S/C6H9NO/c1-2-7-5-3-4-6(7)8/h2H,1,3-5H2

InChI key

WHNWPMSKXPGLAX-UHFFFAOYSA-N

General description

Polyvinylpyrrolidone or PVP is commonly used surfactant or detergent that is nonionic and highly soluble. Ideal for solubilizing and stabilizing proteins, enzymes, and other biomolecules. Ideal for isolation, purification, and characterization. PVP can also be used as a stabilizer to prevent the aggregation or denaturation of proteins during experimental procedures. Its mild nonionic nature and high solubility make it a favorable choice in research applications.

Features and Benefits

  • Redi-Dri™ packaging controls moisture for hygroscopic chemicals that commonly clump
  • Anhydrous powder remains free-flowing and easy to use
  • Safer to handle and weigh out for routine use in lab
  • Eliminates clumps, maintaining product moisture, quality, and attributes

Other Notes

For additional information on our range of Biochemicals, please complete this form.
Polyvinylpyrrolidone is a component of Denhardt′s Solution and is included at a concentration of 1% (w/v) in the standard 50X stock solution.

Legal Information

Redi-Dri is a trademark of Sigma-Aldrich Co. LLC

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
8142081440856568
description

free-flowing, Redi-Dri™, powder, average mol wt 40,000

description

K 30

description

K 90

description

powder, average Mw ~55,000

form

powder

form

powder

form

powder or crystals (or flakes)

form

powder

mol wt

average mol wt 40,000

mol wt

-

mol wt

-

mol wt

average Mw ~55,000

Quality Level

100

Quality Level

200

Quality Level

200

Quality Level

200

product line

Redi-Dri™

product line

-

product line

-

product line

-


Still not finding the right product?

Explore all of our products under Polyvinylpyrrolidone


Storage Class

11 - Combustible Solids

WGK

WGK 1

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters