Skip to Content
Merck
All Photos(1)

Key Documents

R0200000

Reserpine

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

(3β, 16β, 17α, 18β, 20α)-11,17-Dimethoxy-18-[(3,4,5-trimethoxybenzoyl)oxy]yohimban-16-carboxylic acid methyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C33H40N2O9
CAS Number:
Molecular Weight:
608.68
Beilstein:
102014
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

reserpine

manufacturer/tradename

EDQM

mp

~265 °C (dec.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

CO[C@H]1[C@@H](C[C@@H]2CN3CCc4c([nH]c5cc(OC)ccc45)[C@H]3C[C@@H]2[C@@H]1C(=O)OC)OC(=O)c6cc(OC)c(OC)c(OC)c6

InChI

1S/C33H40N2O9/c1-38-19-7-8-20-21-9-10-35-16-18-13-27(44-32(36)17-11-25(39-2)30(41-4)26(12-17)40-3)31(42-5)28(33(37)43-6)22(18)15-24(35)29(21)34-23(20)14-19/h7-8,11-12,14,18,22,24,27-28,31,34H,9-10,13,15-16H2,1-6H3/t18-,22+,24-,27-,28+,31+/m1/s1

InChI key

QEVHRUUCFGRFIF-MDEJGZGSSA-N

Gene Information

human ... SLC18A2(6571)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Reserpine EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Inhibits vesicular uptake of catecholamines and serotonin.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Product No.
Description
Pricing

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Repr. 1A - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Reserpine-induced supersensitivity to the cardiac effects of agonists.
K D Meisheri et al.
Life sciences, 24(6), 473-480 (1979-02-05)
Sandy D Shamon et al.
The Cochrane database of systematic reviews, (4)(4), CD007655-CD007655 (2009-10-13)
Many antihypertensive agents exist today for the treatment of primary hypertension (systolic blood pressure >/=140 mmHg and/or diastolic blood pressure >/=90 mmHg). Randomised controlled trials have been carried out to investigate the evidence for these agents.There is, for example, strong
R E Stitzel
Pharmacological reviews, 28(3), 179-208 (1976-09-01)
Orally administered reserpine is readily absorbed from the GI tract. During this process at least a portion of the drug is metabolized by the intestinal mucosa and then presumably is acted upon by serum esterases. Methylreserpate and trimethoxybenzoic acid are
How reserpine and chlorpromazine act: the impact of key discoveries on the history of psychopharmacology.
G Curzon
Trends in pharmacological sciences, 11(2), 61-63 (1990-02-01)
Reserpine: a challenge for total synthesis of natural products.
Fen-Er Chen et al.
Chemical reviews, 105(12), 4671-4706 (2005-12-15)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service