Skip to Content
Merck

Skip To

PPB005

Phosphate Buffered Saline

powder, pH 7.4, with 0.05% Tween® 20, pHast Pack

Synonym(s):

PBS + 0.05% TWEEN® 20, PBS with 0.05% Tween 20, PBS-T, PBST

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

UNSPSC Code:
12161700
NACRES:
NA.21
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Phosphate Buffered Saline with 0.05% TWEEN® 20, pH 7.4, pHast Pack, powder

product line

pHast Pack

Quality Segment

form

powder

storage condition

(Tightly closed. Dry)

impurities

≤5 ppm Heavy Metals (lead)

color

white

pH

7.2—7.6 (25 °C)

solubility

soluble, clear to hazy, colorless

cation traces

Fe: <10 ppm

foreign activity

DNase, none detected, Endonuclease, none detected, Exonuclease, none detected, NICKase, none detected, Protease, none detected, RNase, none detected

storage temp.

room temp

General description

Phosphate-buffered saline with 0.05% Tween® 20 or PBST contains PBS buffer with a low-concentration of Tween detergent. It is a common wash buffer and due to the presence of detergent, it can easily remove or block excess components thus reducing foaming and non-specific binding on the membrane and microtiter plate wells.

Application

PBST is used in research in a variety of assays including DNA or RNA hybridization assays and cell culture. It is commonly used for immunoassays like Western blot, ELISA, Dot blot, Immunohistochemistry (IHC), immunopercipitation (IP), and immunofluorecence. It has been used for incubation and wash buffer for peptide microarrays for performing High-density peptide array (HDPA) to analyze antibody responses against SARS-CoV-2.

Features and Benefits

  • Ready-to-use buffer
  • No measuring and pH adjusting needed
  • Save time and minimize effort
  • Biological tests: free of DNase, RNase, protease, and nickase
  • Chemical tests: Iron ≤10 ppm, lead ≤5 ppm

Packaging

Foil pouches

Preparation Note

Contents of one pouch, when dissolved in 500 mL of distilled or deionized water, will yield a 1× solution containing 10 mM Na2HPO4, 1.8 mM KH2PO4, 138 mM NaCl, 2.7 mM KCl; 0.05% Tween 20 (w/v), pH 7.4 at 25 °C. Certified to be DNAse, RNAse, Protease, and Nickase free.
Store at room temperature. Product may naturally agglomerate but can be simply broken up within the pouch prior to use.

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Legal Information

TWEEN is a registered trademark of Croda International PLC
pHast Pack is a trademark of Sigma-Aldrich Co. LLC

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
P3563PPB00679378
product line

pHast Pack

product line

-

product line

pHast Pack

product line

BioUltra

Quality Level

100

Quality Level

200

Quality Level

-

Quality Level

200

form

powder

form

powder

form

powder

form

solution

storage temp.

room temp

storage temp.

-

storage temp.

room temp

storage temp.

-

storage condition

(Tightly closed. Dry)

storage condition

-

storage condition

(Tightly closed. Dry)

storage condition

-

solubility

soluble, clear to hazy, colorless

solubility

water: soluble, clear, colorless (1 pouch dissolved in 1L)

solubility

soluble, clear, colorless

solubility

-


Still not finding the right product?


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library







Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/418/501/product-dating-information-06-25-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters