Skip to Content
Merck
All Photos(1)

Documents

PHR1583

Supelco

Megestrol Acetate

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

Megestrol acetate, 17α-Acetoxy-6-methyl-4,6-pregnadiene-3,20-dione, 17α-Hydroxy-6-methyl-4,6-pregnadiene-3,20-dione 17-acetate, 17α-Hydroxy-6-methyl-6-dehydroprogesterone 17-acetate, Megestrol 17-acetate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H32O4
CAS Number:
Molecular Weight:
384.51
Beilstein:
1917291
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to BP 223
traceable to Ph. Eur. Y0001514
traceable to USP 1379106

API family

megesterol

CofA

current certificate can be downloaded

packaging

pkg of 1 g

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

[H][C@]12CC[C@@]3(C)[C@@]([H])(CC[C@]3(OC(C)=O)C(C)=O)[C@]1([H])C=C(C)C4=CC(=O)CC[C@]24C

InChI

1S/C24H32O4/c1-14-12-18-19(22(4)9-6-17(27)13-21(14)22)7-10-23(5)20(18)8-11-24(23,15(2)25)28-16(3)26/h12-13,18-20H,6-11H2,1-5H3/t18-,19+,20+,22-,23+,24+/m1/s1

InChI key

RQZAXGRLVPAYTJ-GQFGMJRRSA-N

Gene Information

human ... PGR(5241)

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards. Megestrol Acetate is a progestogen drug indicated for the palliative treatment of endometrial carcinoma and breast cancer. The administration of the drug is usually done orally as a solid dosage form.

Application

Megestrol Acetate may be used as a pharmaceutical reference standard for the determination of the analyte in pharmaceutical formulations, oral contraceptives and suspensions by chromatography techniques.
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAA4562 in the slot below. This is an example certificate only and may not be the lot that you receive.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Improved bioavailability of a micronized megestrol acetate tablet formulation in humans
Farinha A, et al.
Drug Development and Industrial Pharmacy, 26(5), 567-570 (2000)
Jianghuan Hua et al.
Ecotoxicology and environmental safety, 163, 289-297 (2018-07-30)
Synthetic progestins are widely used pharmaceutical agents that have become common contaminants in the aquatic environment. The potential adverse effects of long-term exposure on aquatic wildlife, however, are not fully understood. The aim of this study was to investigate the
Food effect on the bioavailability of two distinct formulations of megestrol acetate oral suspension
Deschamps B, et al.
International journal of nanomedicine, 4, 185-185 (2009)
Thin-layer chromatographic identification of estrogens and progestogens in oral contraceptives
Simard MB and Lodge BA
Journal of Chromatography A, 51, 517-524 (1970)
Medroxyprogesterone Acetate
USP42-NF37
United States Pharmacopeia/National Formulary, 42(6), 2715-2715 (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service