Skip to Content
Merck
All Photos(1)

Key Documents

PHR1491

Supelco

Neomycin Sulfate

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

Neomycin sulphate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H46N6O13 · xH2SO4
CAS Number:
Molecular Weight:
614.64 (free base basis)
EC Number:
UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to Ph. Eur. N0400000
traceable to Ph. Eur. N0401000
traceable to USP 1458009

API family

neomycin

CofA

current certificate can be downloaded

packaging

pkg of 1 g

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

-10 to -25°C

SMILES string

[S](=O)(=O)(O)O.N[C@H]1[C@H](O[C@@H]([C@H]([C@@H]1O)O)CN)O[C@H]2[C@@H]([C@H]([C@@H](C[C@@H]2N)N)O)O[C@@H]3O[C@@H]([C@H]([C@H]3O)OC4O[C@H]([C@H]([C@@H]([C@H]4N)O)O)CN)CO

InChI

1S/C23H46N6O13.H2O4S/c24-2-7-13(32)15(34)10(28)21(37-7)40-18-6(27)1-5(26)12(31)20(18)42-23-17(36)19(9(4-30)39-23)41-22-11(29)16(35)14(33)8(3-25)38-22;1-5(2,3)4/h5-23,30-36H,1-4,24-29H2;(H2,1,2,3,4)/t5-,6+,7-,8+,9-,10-,11-,12+,13-,14-,15-,16-,17-,18-,19-,20-,21-,22?,23+;/m1./s1

InChI key

OIXVKQDWLFHVGR-GQTDVWSESA-N

Looking for similar products? Visit Product Comparison Guide

General description

Neomycin sulfate is an aminoglycoside broad spectrum antibiotic produced by the fermentation of Streptomyces fradiae or Streptomyces decaris.[1]
Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Application

These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.
Neomycin sulfate may be used as a pharmaceutical reference standard for the determination of the analyte in eye ointments by thin layer chromatography.[2]

Analysis Note

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAC2086 in the slot below. This is an example certificate only and may not be the lot that you receive.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Analysis of neomycin sulfate and framycetin sulfate by high-performance liquid chromatography using evaporative light scattering detection.
Clarot I, et al.
Journal of Chromatography A, 1087(1-2), 236-244 (2005)
Simultaneous identification and quantitative determination of neomycin sulfate, polymixin B sulfate, zinc bacytracin and methyl and propyl hydroxybenzoates in ophthalmic ointment by TLC.
Krzek J, et al.
Journal of Pharmaceutical and Biomedical Analysis, 24(4), 629-636 (2001)
Siddhi Gupta et al.
Antimicrobial agents and chemotherapy, 63(11) (2019-08-21)
Interkingdom polymicrobial biofilms formed by Gram-positive Staphylococcus aureus and Candida albicans pose serious threats of chronic systemic infections due to the absence of any common therapeutic target for their elimination. Herein, we present the structure-activity relationship (SAR) of membrane-targeting cholic
Lokender Kumar et al.
Frontiers in microbiology, 12, 598498-598498 (2021-02-16)
Pseudomonas aeruginosa utilizes the quorum sensing (QS) system to strategically coordinate virulence and biofilm formation. Targeting QS pathways may be a potential anti-infective approach to treat P. aeruginosa infections. In the present study, we define cephalosporins' anti-QS activity using Chromobacterium

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service