Skip to Content
Merck
All Photos(1)

Key Documents

PHR1326

Supelco

Calcium carbonate

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

Calcium carbonate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CaCO3
CAS Number:
Molecular Weight:
100.09
Beilstein:
8008338
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to USP 1086403

API family

calcium carbonate

CofA

current certificate can be downloaded

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

density

2.93 g/mL at 25 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-30°C

SMILES string

[Ca++].[O-]C([O-])=O

InChI

1S/CH2O3.Ca/c2-1(3)4;/h(H2,2,3,4);/q;+2/p-2

InChI key

VTYYLEPIZMXCLO-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards. Calcium carbonate is a material that is widely found in nature, and is commonly referred to as calcite. Calcium carbonate is used in such processes as the manufacture of paper, paints, glass, and ceramics, the compounding of polymers, environmental applications such as wastewater treatment, and as an extender in paints and coatings.

Application

These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAC3446 in the slot below. This is an example certificate only and may not be the lot that you receive.

related product

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Calcium carbonate urolithiasis in an adult male cynomolgus monkey.
C M O'Rourke et al.
Laboratory animal science, 45(2), 222-224 (1995-04-01)
Dongni Ren et al.
Micron (Oxford, England : 1993), 42(3), 228-245 (2010-10-19)
The review focuses on the effects of several important additives and templates controlling the calcium carbonate crystals formation and the complexity of the crystal morphologies in vitro. Additives include soluble matrices extracted from shells and pearls, amino-acids, magnesium ions and
Julyan H E Cartwright et al.
Angewandte Chemie (International ed. in English), 51(48), 11960-11970 (2012-11-06)
Although the polymorphism of calcium carbonate is well known, and its polymorphs--calcite, aragonite, and vaterite--have been highly studied in the context of biomineralization, polyamorphism is a much more recently discovered phenomenon, and the existence of more than one amorphous phase
Liliana Astachov et al.
Frontiers in bioscience (Landmark edition), 16, 458-471 (2011-01-05)
Stem cell development and fate decisions are dictated by the microenvironment in which the stem cell is embedded. Among the advanced goals of tissue engineering is the creation of a microenvironment that will support the maintenance and differentiation of the
Polysaccharides and proteoglycans in calcium carbonate-based biomineralization.
José L Arias et al.
Chemical reviews, 108(11), 4475-4482 (2008-07-26)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service