Skip to Content
Merck
All Photos(1)

Key Documents

PHR1179

Supelco

Chlorothiazide

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

6-Chloro-2H-1,2,4-benzothiadiazine-7-sulfonamide 1,1-dioxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H6ClN3O4S2
CAS Number:
Molecular Weight:
295.72
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to BP 76
traceable to Ph. Eur. C1700000
traceable to USP 1121005

API family

chlorothiazide

CofA

current certificate can be downloaded

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

O=S(C1=C2C=C(Cl)C(S(N)(=O)=O)=C1)(NC=N2)=O

InChI

1S/C7H6ClN3O4S2/c8-4-1-5-7(2-6(4)16(9,12)13)17(14,15)11-3-10-5/h1-3H,(H,10,11)(H2,9,12,13)

InChI key

JBMKAUGHUNFTOL-UHFFFAOYSA-N

Gene Information

human ... SLC12A3(6559)

Looking for similar products? Visit Product Comparison Guide

General description

Chlorothiazide belongs to the class of thiazide diuretic drugs. It is used for the management of postparturient udder edema in cattle. Its mode of action involves the inhibition of chloride reabsorption in the proximal portion of distal tubule.
Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Application

Chlorothiazide may be used as a pharmaceutical reference standard for the determination of the analyte in pharmaceutical formulations by chromatography.
One of the first in the thiazide class of diuretics.
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAA8998 in the slot below. This is an example certificate only and may not be the lot that you receive.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chlorothiazide
USP42-NF37: United States Pharmacopeia and National Formulary
United States Pharmacopeia/National Formulary, 29(5), 942-942 (2019)
Pharmacologic Adjuncts I
Assisted Ventilation of the Neonate, 311-328 (2003)
Liquid chromatography in pharmaceutical analysis II: Determination of a reserpine?chlorothiazide mixture
Honigberg IL, et al.
Journal of Pharmaceutical Sciences, 63(11), 1762-1764 (1974)
Validated micellar electrokinetic capillary chromatography method for quality control of the drug substances hydrochlorothiazide and chlorothiazide
Thomas BR, et al.
Journal of Chromatography. B, Biomedical Sciences and Applications, 657(2), 383-394 (1994)
Liquid chromatographic determination of chlorothiazide and hydrochlorothiazide diuretic drugs in bovine milk
Shaikh B and Rummel N
Journal of Agricultural and Food Chemistry, 46(3), 1039-1043 (1998)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service