Skip to Content
Merck
All Photos(1)

Key Documents

PHR1173

Supelco

N-Acetyl-L-tyrosine

Pharmaceutical Secondary Standard; Certified Reference Material

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
4-(HO)C6H4CH2CH(NHCOCH3)CO2H
CAS Number:
Molecular Weight:
223.23
Beilstein:
2697172
EC Number:
MDL number:
UNSPSC Code:
41116107
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to USP 1010106

API family

acetyltyrosine

CofA

current certificate can be downloaded

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

149-152 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

CC(=O)N[C@@H](Cc1ccc(O)cc1)C(O)=O

InChI

1S/C11H13NO4/c1-7(13)12-10(11(15)16)6-8-2-4-9(14)5-3-8/h2-5,10,14H,6H2,1H3,(H,12,13)(H,15,16)/t10-/m0/s1

InChI key

CAHKINHBCWCHCF-JTQLQIEISA-N

Looking for similar products? Visit Product Comparison Guide

General description

N-Acetyl-L-tyrosine is an acetyl derivative of the amino acid, L-tyrosine. It is freely soluble in water compared to L-tyrosine and is commonly used as a parenteral nutrition supplement.
Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Application

N-Acetyl-L-tyrosine may be used as a pharmaceutical reference standard for the determination of the analyte in bulk drug and pharmaceutical formulations by titrimetry and high performance liquid chromatography.
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.
Values of analytes vary lot to lot.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAA6108 in the slot below. This is an example certificate only and may not be the lot that you receive.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

N-Acetyltyrosine
European Pharmacopoeia Commission and European Directorate for the Quality of Medicines & Healthcare
European pharmacopoeia, 1641-1642 (2017)
N-Acetyltyrosine
Pharmacopeia, US
United States Pharmacopeia/National Formulary, 38(3)(5), 4702-4702 (2018)
Rakesh Kumar et al.
European journal of medicinal chemistry, 42(4), 503-510 (2006-12-26)
The development of type 2 diabetes in obese individuals is linked to lipid accumulation in non-adipose tissues. A series of N-acetyl-L-tyrosine derivatives were synthesized and evaluated for PPAR transactivation. Compounds 4d and 4f were found to show better PPARalpha transactivation
Wei Xie et al.
Journal of inorganic biochemistry, 103(1), 58-63 (2008-10-25)
Complexes of the uranyl cation [UO(2)(2+)] with histidine (His), N-acetyl-histidine (NAH), tyrosine (Tyr), and N-acetyl-tyrosine (NAT) were studied by UV-visible and NMR spectroscopy, and by potentiometric titration. Protonation constants for each ligand are reported, as are cumulative formation constants for
David I Pattison et al.
Biochemistry, 45(26), 8152-8162 (2006-06-28)
Hypochlorous acid (HOCl) is a powerful oxidant generated from H(2)O(2) and Cl(-) by the heme enzyme myeloperoxidase, which is released from activated leukocytes. HOCl possesses potent antibacterial properties, but excessive production can lead to host tissue damage that is implicated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service