Skip to Content
Merck
All Photos(1)

Key Documents

PHR1119

Supelco

D-Camphor

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

2-Bornanone, 2-Camphanone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H16O
CAS Number:
Molecular Weight:
152.23
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to Ph. Eur. C0405000
traceable to USP 1087508

vapor density

5.24 (vs air)

vapor pressure

4 mmHg ( 70 °C)

API family

camphor

CofA

current certificate can be downloaded

autoignition temp.

870 °F

expl. lim.

3.5 %

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

178-182 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

CC1(C)[C@@H]2CC[C@@]1(C)C(=O)C2

InChI

1S/C10H16O/c1-9(2)7-4-5-10(9,3)8(11)6-7/h7H,4-6H2,1-3H3/t7-,10+/m1/s1

InChI key

DSSYKIVIOFKYAU-XCBNKYQSSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards. D-Camphor is an acyclic terpenoid and one of the key volatile constituents of essential oils of plants Artemisia tridenta, Mentha longifolia ssp., Longifolia and some Lamiaceae species.

Application

These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAA8982 in the slot below. This is an example certificate only and may not be the lot that you receive.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Eye Dam. 1 - Flam. Sol. 2 - Skin Irrit. 2 - STOT SE 2 Inhalation

Target Organs

Lungs

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 1

Flash Point(F)

147.2 °F - closed cup

Flash Point(C)

64 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Jeffrey N Love et al.
The Journal of emergency medicine, 27(1), 49-54 (2004-06-29)
Serious pediatric toxicity resulting from exposure to small amounts of camphor-containing products has long been a problem. Twenty years ago the United States Food and Drug Administration took several actions in an attempt to ameliorate this risk. Despite these changes
E Siegel et al.
Pediatric clinics of North America, 33(2), 375-379 (1986-04-01)
Camphor is present in several over-the-counter compounds of questionable use and therefore may be ingested by small children. Because seizures may follow ingestion of certain amounts, appropriate treatment is needed, including the use of anticonvulsants.
Weiyang Chen et al.
Molecules (Basel, Switzerland), 18(5), 5434-5454 (2013-05-15)
The fragrant camphor tree (Cinnamomum camphora) and its products, such as camphor oil, have been coveted since ancient times. Having a rich history of traditional use, it was particularly used as a fumigant during the era of the Black Death
Camphor: a chiral starting material in natural product synthesis.
T Money
Natural product reports, 2(3), 253-289 (1985-06-01)
Gustav Georg Belz et al.
Phytomedicine : international journal of phytotherapy and phytopharmacology, 10 Suppl 4, 61-67 (2003-06-17)
Independent, double-blinded, randomized, placebo-controlled studies using sublingual/oral administration of D-camphor, an extract from fresh crataegus berries, and a combination of the two (CCC) yielded the following results: Both the D-camphor and the extract from fresh crataegus berries, the components of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service