Skip to Content
Merck
All Photos(1)

Key Documents

PHR1078

Supelco

Phytonadione

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

Vitamin K1, Phytomenadione, 2-Methyl-3-phytyl-1,4-naphthoquinone, 3-Phytylmenadione, Phylloquinone, Vitamin K1(20)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C31H46O2
CAS Number:
Molecular Weight:
450.70
Beilstein:
2568816
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to BP 423
traceable to Ph. Eur. P1609000
traceable to USP 1538006

API family

phytonadione

CofA

current certificate can be downloaded

storage condition

protect from light

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.527 (lit.)

mp

−20 °C (lit.)

density

0.984 g/mL at 25 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

O=C(C1=CC=CC=C21)C(C)=C(C/C=C(CCC[C@@H](CCC[C@H](C)CCCC(C)C)C)\C)C2=O

InChI

1S/C31H46O2/c1-22(2)12-9-13-23(3)14-10-15-24(4)16-11-17-25(5)20-21-27-26(6)30(32)28-18-7-8-19-29(28)31(27)33/h7-8,18-20,22-24H,9-17,21H2,1-6H3/b25-20+/t23-,24-/m1/s1

InChI key

MBWXNTAXLNYFJB-NKFFZRIASA-N

Looking for similar products? Visit Product Comparison Guide

General description

Phytonadione belongs to the category of oral anticonvulsant drugs. It protects a newborn from acquiring hemorrhagic diseases by maintaining a normal blood clotting mechanism.[1][2]
Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Application

Phytonadione may be used as a pharmaceutical reference standard in the determination of the analyte in pharmaceutical formulations by chromatographic[3] and spectrophotometric techniques.[4]
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.
Values of analytes vary lot to lot.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAB3714 in the slot below. This is an example certificate only and may not be the lot that you receive.

related product

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Comparison of oral vs intravenous phytonadione (vitamin K1) in patients with excessive anticoagulation: a prospective randomized controlled study
Lubetsky A, et al.
Archives of Internal Medicine, 163(20), 2469-2473 (2003)
Treatment of excessive anticoagulation with phytonadione (vitamin K): a meta-analysis
DeZee KJ, et al.
Archives of Internal Medicine, 166(4), 391-397 (2006)
NP-HPLC method for determining phytonadione and its impurities
Xue M, et al.
Journal of Liquid Chromatography and Related Technologies, 39(17-18), 799-805 (2016)
Development and validation of spectrophotometric methods fordetermination of phytomenadione in injection
Patel SA and Parikh PK
Current Research in Pharmaceutical Sciences, 39(17-18), 26-30 (2012)
M Kyla Shea et al.
The American journal of clinical nutrition, 98(1), 197-208 (2013-05-31)
Animal studies have shown that vitamin K treatment reduced vascular calcification, but human data are limited. We determined the association between vitamin K status and coronary artery calcium (CAC) progression in the Multi-Ethnic Study of Atherosclerosis by using a case-cohort

Questions

1–3 of 3 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  3. Could you clarify certified purity calculation for batch LRAD1021? CoA says 98.6% but I find 99.2% when using the formula and test results provided.

    1 answer
    1. The Loss On Drying (LOD) value is not provided on the Certificate of Analysis. This value was used in the final purity calculation to arrive at 98.6% the reported purity value. If not corrected for LOD, you will get a result of 99.2%.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service