Skip to Content
Merck
All Photos(1)

Key Documents

NIST772A

Magnetic moment standard - nickel sphere

NIST® SRM® 772a

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

form

spheres

packaging

pkg of each

manufacturer/tradename

NIST®

application(s)

general analytical

General description

This Standard Reference Material (SRM) is intended for use in the calibration of magnetometers (such as vibrating sample magnetometers) used in the measurement of the magnetic properties of materials. SRM 762 consists of a nickel disk nominally 6 mm in diameter with a thickness of 0.127 mm and a mass of 32 mg ± 1 mg. For more information, please refer to the COA and SDS.

SRM 772A_cert

SRM 772A_SDS

Other Notes

Example analytes are listed below as a reference. Please download a current certificate at nist.gov/SRM for current analytes and certified values.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 2 - Skin Sens. 1 - STOT RE 1

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


  • Choose from one of the most recent versions:

    Certificates of Analysis (COA)

    Lot/Batch Number

    Sorry, we don't have COAs for this product available online at this time.

    If you need assistance, please contact Customer Support.

    Already Own This Product?

    Find documentation for the products that you have recently purchased in the Document Library.

    Visit the Document Library

    Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

    Contact Technical Service