Skip to Content
Merck
All Photos(1)

Key Documents

NIST350C

Benzoic acid

NIST® [TM′"SRM"] 350c

Synonym(s):

Benzenecarboxylic acid, Benzeneformic acid, Benzenemethanoic acid, Benzoate; carboxybenzene, C6H5COOH, C7H6O2, Dracylic acid, Phenyl carboxylic acid, Phenylcarboxylic acid, Phenylformic acid

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

form

crystals

packaging

pkg of 30 g

manufacturer/tradename

NIST®

technique(s)

titration: suitable

General description

This SRM350c Standard Reference Material (SRM) consists of purified benzoic acid. NIST-certified mass fraction value of the SRM is equal to 99.959% ± 0.040%.

SRM 350C_cert

SRM 350C_SDS

Application

Benzoic acid NIST standard is intended for use in acidimetric standardization.

Features and Benefits

  • Certified mass fraction value is traceable to SI units for mass, current, and time via coulometric assay.
  • Details of expiration date, homogeneity, and storage and use instructions are provided on NIST certificate.
  • A unit of SRM 350c consists of 30 g of benzoic acid.

Other Notes

Certified for the analytes listed below.
Benzoic Acid (C6H5COOH)

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

related product

Product No.
Description
Pricing

Pictograms

Health hazardCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2 - STOT RE 1 Inhalation

Target Organs

Lungs

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 1

Flash Point(F)

249.8 °F - closed cup

Flash Point(C)

121 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service